Product Image
Applied Biosystems™
SeqStudio™ Genetic Analyzer for HID Install Kit (sequencing & fragment analysis)
This install kit is for use with the SeqStudio Genetic Analyzer for HID. It includes all of the consumables andRead more
Have Questions?
Catalog NumberQuantity
A461801 Kit
Catalog number A46180
Price (INR)
532,710.00
Each
Quantity:
1 Kit
Price (INR)
532,710.00
Each
This install kit is for use with the SeqStudio Genetic Analyzer for HID. It includes all of the consumables and accessories needed to perform an instrument qualification (install, operational, or/and performance). This kit supports both fragment analysis and sequencing applications. For an install kit that supports fragment analysis only, please see Cat. No. A46182.
Specifications
For Use With (Application)Sequencing, Human Identification
For Use With (Equipment)Sequencing Instruments (Capillary Electrophoresis-Based)
Product LineSeqStudio
Product TypeGenetic Analyzer
Quantity1 Kit
Shipping ConditionDry Ice
SpeciesHuman
FormatKit
Unit SizeEach