SeqStudio™ Genetic Analyzer for HID, desktop, with training and 1-year extra warranty
Applied Biosystems™

SeqStudio™ Genetic Analyzer for HID, desktop, with training and 1-year extra warranty

The Applied Biosystems SeqStudio Genetic Analyzer for HID is a low-throughput, easy-to-use, convenient benchtop system that delivers gold-standard fragment analysisRead more
Have Questions?
Catalog NumberQuantity
A463681 instrument package
Catalog number A46368
Price (INR)
-
Quantity:
1 instrument package
The Applied Biosystems SeqStudio Genetic Analyzer for HID is a low-throughput, easy-to-use, convenient benchtop system that delivers gold-standard fragment analysis or Sanger sequencing with just a simple click. It is validated across a broad range of HID forensic kits to deliver answers you can trust. The SeqStudio Genetic Analyzer couples the latest advancements in touchscreen usability with the same data quality, reliability, and support you have come to expect from Applied Biosystems instruments.

Features of the SeqStudio Genetic Analyzer for HID include:
• Universal all-in-one cartridge—integrates POP-1 polymer, anode buffer, a polymer delivery system, and a 4-capillary array to minimize instrument setup and maintenance time; allows for on-instrument reagent life of up to six months and 250 injections (1000 samples)
• Trusted results—optimized data collection software (SeqStudio DCS v1.2) and validation certificates for use with many Applied Biosystems STR kits
• SAE software—security, audit, and e-signature functionality for protection and traceability
• Compact design—configurable with either a laptop or desktop computer; the smallest footprint of all of our genetic analyzer systems
• Easy operation—auto-calibration with RFID enabled tracking of consumables usage and interactive touch-screen controls for streamlined workflow
• World-class training and support—every SeqStudio analyzer for HID includes a one-day training by our forensic FAS team to get you up and running quickly; subsequent support by our >2800 field service engineers and technical support scientists located around the globe

The SeqStudio Genetic Analyzer employs a cartridge-based system that is easy to use and easy to maintain. The all-in-one cartridge contains the capillary array, polymer reservoir, and anode buffer. The cartridge design allows for up to six months of on-instrument reagent storage, with a universal polymer (POP-1 polymer) that is compatible with both Sanger sequencing and fragment analysis. The SeqStudio analyzer is compatible with standard 96-well plates or 8-well strip tubes and can generate results in less than 40 minutes per run.

Validation
The SeqStudio Genetic Analyzer for HID and DCS v1.2 software were validated according to the Scientific Working Group for DNA Analysis Methods (SWGDAM, December, 2016) for use with the following kits: GlobalFiler, GlobalFiler Express, VeriFiler Plus, VeriFiler Express, Huaxia Platinum, Identifiler Plus, MiniFiler, NGMSelect, NGM Detect, YFiler, YFiler Plus PCR amplification kits. Details and data from the studies, including sizing precision and accuracy, genotyping concordance, resolution, sensitivity and dynamic range, minimum detection threshold, and contamination and mixtures studies, can be found in the SeqStudio Validation User Bulletin. Each laboratory should perform internal validation studies to establish appropriate guidelines, protocols, and procedures prior to implementing a new technology.

Training
Included training is conducted at the customer site by an HID Field Application Scientist. The one-day training includes both classroom instruction as well as a hands-on review of the instrument and user interface. Optionally, a wet-lab section can be accommodated with a walk-through of run setup and performance of an actual injection.

Warranty
The included extended warranty plan adds additional coverage to the existing manufacturer’s limited warranty. The extended warranty includes a planned maintenance visit. Should your instrument require a repair, the extended warranty coverage includes parts, labor, and travel at no additional cost. In addition, you’ll receive:

• On-site guaranteed 2-day response time*
• Scheduled on-site planned maintenance
• Priority access to remote service engineers

Certain exclusions apply. Contact your Services and Support Representative for details at service.sales@thermofisher.com.

Notes:
• SeqStudio HID Install kits (Cat. Nos. A46190 or A46182), SeqStudio Cartridge (A41328), GeneMapper IDx software, [extended warranty,] and other system consumables and reagents sold separately.
• A computer is required to run the SeqStudio analyzer for HID with SAE software, so for new installations, we recommend purchasing package A46228 or A46229. For multiple SeqStudio analyzer purchases or other installation configurations, please consult with your local sales representative.

*Availability limited in some geographical areas.
Specifications
For Use With (Equipment)Sequencing Instruments (Capillary Electrophoresis-Based)
Format96-well Plate, 8-tube Strip
Quantity1 instrument package
Shipping ConditionRoom Temperature
Product LineSeqStudio
Software TypeData Collection Software
Unit SizeEach
Package Components
A36187
Catalog Number
A36187
Unit Size
Qty
1
A46208
Catalog Number
A46208
Unit Size
Qty
1
A34272
Catalog Number
A34272
Unit Size
Qty
1
ZG61SCSEQSTUDIO
Catalog Number
ZG61SCSEQSTUDIO
Unit Size
Qty
1

Frequently asked questions (FAQs)

How do I set up the same user, system security, audit, and e-signature settings on multiple instances of the SAE Administrator Console v2.0?

You may use the export and import functions to transfer settings from one installation of the SAE Administrator Console to another.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

What does archiving audit records in the SAE Administrator Console do?

Archiving audit records removes the records from the SAE Administrator Console and saves them in an internally specified location on the same computer on which the SAE Administrator Console is installed. Note: Archived audit records are accessible for viewing in the SAE Administrator Console.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

What actions are auditable in the SAE Administrator Console v2.0?

Auditable actions in the SAE Administrator Console are:
- Enable or disable security, audit, or e-signature
- Sign in to or out of the SAE Administrator Console
- Import or export an SAE configuration
- Install an application profile
- Archive, purge, or restore audit records
- Manual sync with LDAP (Lightweight Directory Access Protocol) Directory

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

Can I enable or disable the system security function when using the SAE Administrator Console v2.0?

The system security function cannot be disabled in the SAE Administrator Console. To disable user sign in to an application, you must disable SAE in the application.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.

What happens if I forget my password when using the SAE Administrator Console v2.0?

There is no way to recover a forgotten password. If the SAE administrator forgets their password, the software must be reinstalled. Export all data before reinstalling the software.

Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.