Acetic Acid, Glacial (Optima™), Fisher Chemical™
Acetic Acid, Glacial (Optima™), Fisher Chemical™
Acetic Acid, Glacial (Optima™), Fisher Chemical™
Fisher Chemicals

Acetic Acid, Glacial (Optima™), Fisher Chemical™

Have Questions?
Catalog NumberQuantityPackaging
A465-11 LFEP Bottle
Catalog number A465-1
HSN 29152100
Price (INR)
225,212.00
Each
Quantity:
1 L
Packaging:
FEP Bottle
Request bulk or custom format
Price (INR)
225,212.00
Each
Specifications
Chemical Name or MaterialAcetic Acid
Cleanup SymbolAcid
pH2.5
CAS64-19-7
ChemAlert Storage SymbolRed
View more
  • Certificate of Analysis included with each bottle.
  • Ideal for analysis by ICP-MS and high-resolution ICP-MS
  • Evaluated for 65 metals at low-level ppt concentrations.
  • Manufactured, purified, and packaged in a Class 1000 clean room.
  • Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA.