GeneArt™ Gibson Assembly HiFi Cloning Kit, chemically competent cells
GeneArt™ Gibson Assembly HiFi Cloning Kit, chemically competent cells
Invitrogen™
GeneArt™ Gibson Assembly HiFi Cloning Kit, chemically competent cells
The GeneArt Gibson Assembly HiFi Cloning Kit enables DNA assembly and cloning via a technique that allows multiple overlapping DNARead more
Have Questions?
Catalog NumberQuantity
A4662410 Reactions
Catalog number A46624
Price (INR)
20,266.00
Each
Add to cart
Quantity:
10 Reactions
Price (INR)
20,266.00
Each
Add to cart
The GeneArt Gibson Assembly HiFi Cloning Kit enables DNA assembly and cloning via a technique that allows multiple overlapping DNA fragments to be seamlessly linked in a one-step, 15–60 minute isothermal reaction. DNA fragments of different lengths are uniformly assembled using complementary overlaps between fragments. The inherent flexibility of this approach is suitable for small and large DNA constructs and includes both single and multiple inserts. The resulting products can be used for a variety of downstream applications including transformation, PCR and rolling circle amplification (RCA). The GeneArt Gibson Assembly HiFi Cloning Kit, electrocompetent cells, is a complete kit that includes master mix, positive control, water, and One Shot TOP10 chemically competent E. coli.
Features of the GeneArt Gibson Assembly HiFi Cloning Kit include:
Simple—seamlessly assemble and clone up to six DNA fragments in a single reaction
Efficient—high fidelity provides more correct clones than other methods
Flexible—design guidelines allow assembly into any vector of your choice
Convenient—available as master mix kits and cloning kits complete with chemical or electrocompetent cells
Trusted—over 4000 citations in the scientific literature highlighting great success
Seamless assembly of multiple fragments
GeneArt Gibson Assembly HiFi cloning is a simple, one-step process whereby up to six fragments are combined in a proprietary enzymatic mix in order to assemble DNA fragments with shared terminal end homology without leaving any extra sequences or scars behind (seamless). After a 15–60 minute incubation, a portion of the assembly reaction is then transformed into chemically competent or electrocompetent cells, yielding clones that are ready for analysis the next day. The required 20- to 40-base pair end homology is designed into the de novo fragment for synthesis or can be easily engineered by PCR amplification with custom DNA oligos. This special enzyme mix creates a seamless and covalently bound DNA construct providing high efficiency.
Robust method for maximum efficiency
Due to the covalently bound final product, the GeneArt Gibson Assembly HiFi method allows the utilization of chemically competent cells or electrocompetent cells for the highest transformation efficiency, improving the success of finding the right clone, particularly for more challenging constructs. There is no need for restriction enzymes, ligation, or recombination sites, and the method provides a perfectly seamless construct without unwanted extra bases.
Provides great versatility
The GeneArt Gibson Assembly HiFi method provides versatility, can streamline many techniques through the rapid combination, addition, deletion, or exchange of DNA segments, and eliminates the need to subclone, saving time and effort in the cloning workflow.
Convenient formats
GeneArt Gibson Assembly HiFi kits are available in multiple formats, including:
• GeneArt Gibson Assembly HiFi Cloning Kit, chemically competent cells (with One Shot TOP10 chemically competent cells) (this page)
GeneArt Gibson Assembly HiFi Cloning Kit, electrocompetent cells (with ElectroMAX DH10B electrocompetent cells)
GeneArt Gibson Assembly HiFi Master Mix kits (cloning kit without the cells)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeChemically Competent E. coli
For Use With (Application)Cloning
Number of FragmentsUp to 6 fragments
Product LineGENEART
Product TypeCloning Kit
Quantity10 Reactions
Shipping ConditionDry Ice
FormatKit
Unit SizeEach
Contents & Storage
Includes master mix, positive control, water, and One Shot TOP10 chemically competent E. coli.
Frequently asked questions (FAQs)
How big do DNA fragment homology arms need to be when using Gibson Assembly kits?
For fragments that are 8 kb or lesser in length, an overlap of 20-40 bp is recommended. For fragments greater than 8 kb in length, an overlap of 40-100 bp is recommended.
What type of vector can I use with Gibson Assembly kits?
Any linearized vector can be used with Gibson Assembly kits. We recommend using high-copy vectors for small inserts (<10 kb) and low-copy vectors for large inserts or toxic genes.
What type of inserts can I use with Gibson Assembly kits?
Any linearized DNA fragment, including PCR products, restriction digestion products, and GeneArt Strings can be used with Gibson Assembly kits.
What is the difference between the Gibson Assembly HiFi and the Gibson Assembly EX kits?
Gibson Assembly HiFi kits provide the quickest and easiest way to assemble up to six fragments in a single-tube reaction. The more robust Gibson Assembly EX kits enable assembly of up to 15 fragments in a single-tube, two-step reaction.