Product Image
Applied Biosystems™
MicroSEQ™ ID 16S rDNA 500 Microbial Library, v2019
The MicroSEQ ID 16S rDNA 500 Microbial Library v2019 is for use with MicroSEQ ID Microbial Identification Software v3.0 and v3.1. It contains over 2100 reference 16S rRNA sequences from bacteria.
Have Questions?
Catalog NumberQuantity
A471341 Ea.
Catalog number A47134
Price (INR)
1,583,850.00
Each
Quantity:
1 Ea.
Price (INR)
1,583,850.00
Each
The MicroSEQ ID 16S rDNA 500 Microbial Library v2019 is for use with MicroSEQ ID Microbial Identification Software v3.0 and v3.1. It contains over 2100 reference 16S rRNA sequences from bacteria. The library entries contain the first 500 bases of the 16S rRNA gene, enough to identify bacterial unknowns. The library is delivered in either digital or USB format.
Specifications
Product LineMicroSEQ™ ID
Quantity1 Ea.
Shipping ConditionRoom Temperature
Unit SizeEach