AAV-MAX pAAV-Rep2Cap2 Vector
Gibco™

AAV-MAX pAAV-Rep2Cap2 Vector

AAV-MAX pAAV-Rep2Cap2 Vector is one of the three plasmids included in the AAV-MAX Control Plasmids kit (Cat. No. A47672), madeRead more
Have Questions?
Catalog NumberQuantity
A476781 Tube
Catalog number A47678
Price (INR)
70,620.00
Each
Quantity:
1 Tube
Price (INR)
70,620.00
Each
AAV-MAX pAAV-Rep2Cap2 Vector is one of the three plasmids included in the AAV-MAX Control Plasmids kit (Cat. No. A47672), made available here separately. These three plasmids can be used together to produce adeno-associated virus serotype 2 (AAV2)−expressing GFP and consist of pAAV-CMV-emGFP, a transfer plasmid, pAAV-Rep2Cap2, a Rep/Cap plasmid, and pHelper, a helper plasmid. They must be used with a HEK293 cell line that stably expresses the adenovirus E1 gene. Offering the plasmids as standalone products provides users with flexibility to optimize plasmid ratios as desired.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1 mg/mL
Delivery TypeTransfection
Expression SystemMammalian
Product LineAAV-MAX
Product TypePlasmid
Quantity1 Tube
Shelf Life2 Years
Shipping ConditionIce
Regulatory StatusRUO
SterilityNon-sterile
Unit SizeEach
Contents & Storage
• 1 x 0.75 mL tube, 1 mg/mL

Store at -20°C.