Product Image
Applied Biosystems™
384-Well Plate Retainer & Base Set, standard, for SeqStudio™ Flex
This 384-well plate Retainer and Base Set is for use with standard 384-well plates on the SeqStudio 24 Flex Genetic Analyzer.
Have Questions?
Catalog NumberQuantity
A494964 Sets
Catalog number A49496
Price (INR)
27,609.00
Each
Quantity:
4 Sets
Price (INR)
27,609.00
Each
This 384-well plate Retainer and Base Set is for use with standard 384-well plates on the SeqStudio 24 Flex Genetic Analyzer.
This 384-well plate Retainer and Base Set is for use with standard 384-well plates on the SeqStudio 24 Flex Genetic Analyzer.
Specifications
For Use With (Application)Sanger Sequencing and Fragment Analysis
For Use With (Equipment)SeqStudio™ Flex 24-cap Genetic Analyzers
Plate CompatibilityStandard 384-well plates
Quantity4 Sets
Unit SizeEach
Frequently asked questions (FAQs)
When shouldn't I use the 3500 plate retainers on the SeqStudio Flex Series Genetic Analyzers?
If the automatic barcode workflow is enabled on the SeqStudio Flex Series Genetic Analyzer, using the 3500 plate retainers will cause a barcode error in the home screen.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
What is different about the SeqStudio Flex Series Genetic Analyzer plate retainers compared with the 3500 plate retainers?
The SeqStudio Flex Series Genetic Analyzer plate retainers have one opening on the side to allow barcode reading while the 3500 plate retainers have two openings on the side which can cover the plate barcode.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.