Search
| Catalog Number | Quantity |
|---|---|
| A50251 | 96 samples |
The Ion SingleSeq GX 201-296 Kit is designed for low cell number input library construction prior to low-pass genome sequencing detection of copy number events. Requiring as little as 6 pg of DNA from a single cell, or input from multiple cells, this kit includes reagents to extract, amplify, and barcode genomic DNA for preparation of libraries from up to 96 samples.
The Ion SingleSeq GX 201-296 Kit is intended to be used with the Ion ReproSeq PGT-A GX5 Kit and Genexus System for the detection of chromosomal aneuploidies and chromosome arm copy number events (>48 Mb). The Ion ReproSeq PGT-A GX5 Kit is used with the Genexus System to enable automated template preparation, chip loading, and sequencing for fast and simple instrument set-up to minimize hands-on time.
Notes
• This kit is compatible with the Genexus system.