Ion SingleSeq™ GX 301-396 Kit
Ion Torrent™
Ion SingleSeq™ GX 301-396 Kit
The Ion SingleSeq GX 301-396 Kit is designed for low cell number input library construction prior to low-pass genome sequencingRead more
Have Questions?
Catalog NumberQuantity
A5025296 samples
Catalog number A50252
Price (INR)
1,497,696.00
Each
Quantity:
96 samples
Price (INR)
1,497,696.00
Each
The Ion SingleSeq GX 301-396 Kit is designed for low cell number input library construction prior to low-pass genome sequencing detection of copy number events. Requiring as little as 6 pg of DNA from a single cell, or input from multiple cells, this kit includes reagents to extract, amplify, and barcode genomic DNA for preparation of libraries from up to 96 samples.
The Ion SingleSeq GX 301-396 Kit is intended to be used with the Ion ReproSeq PGT-A GX5 Kit and Genexus System for the detection of chromosomal aneuploidies and chromosome arm copy number events (>48 Mb). The Ion ReproSeq PGT-A GX5 Kit is used with the Genexus System to enable automated template preparation, chip loading, and sequencing for fast and simple instrument set-up to minimize hands-on time.
Notes
• This kit is compatible with the Genexus system.
For Research Use Only.
Specifications
Quantity96 samples
Unit SizeEach