Pierce™ High Capacity Ni-IMAC MagBeads, EDTA compatible
Thermo Scientific™
Pierce™ High Capacity Ni-IMAC MagBeads, EDTA compatible
Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads are designed for purification of over-expressed His-tagged proteins from cell culture media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A50591100 mL
A505881 mL
A505895 mL
A5059025 mL
Catalog number A50591
Price (INR)
682,461.00
Each
Quantity:
100 mL
Price (INR)
682,461.00
Each
Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads are designed for purification of over-expressed His-tagged proteins from cell culture media. The beads have high binding capacity (up to 80 mg of protein per mL of settled beads) and can tolerate the high EDTA and reducing agent concentrations that are used to inhibit metalloproteases and stabilize sensitive intracellular proteins.
Features of the high-capacity EDTA-compatible Ni-IMAC MagBeads include:
  • Chelator stable—stable in buffer containing 20 mM EDTA and DTT
  • Compatible with purification of overexpressed, secreted proteins in cell culture media (e.g., Expi293, ExpiCHO, and ExpiSf expression systems)
  • Exceptional high protein binding capacity—up to 80 mg/mL of settled beads
  • High-performance—non-aggregating, superparamagnetic microparticles provide exceptional performance with manual and automated HTS applications (e.g., Thermo Scientific KingFisher instruments)
Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads consist of crosslinked agarose beads embedded with magnetite. The density of the ligand on the magnetic agarose bead results in a binding capacity similar to or better than traditional agarose resins with the added benefits of having magnetic properties and being EDTA compatible. The MagBeads are a valuable tool for small-scale purification of multiple His-tagged proteins and for scouting expression and purification conditions to be used in larger-scale purifications.
Pierce high-capacity EDTA-compatible Ni-IMAC MagBeads are optimized for use with high-throughput magnetic platforms, such as the KingFisher 96 and KingFisher Flex magnetic particle processors, but the beads also compatible with simple benchtop purification applications using an appropriate magnetic stand. Automated instruments, however, are especially useful for higher throughput purification and screening of purification conditions.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bed Volume25 mL
Binding PropertyChelator Stable
Column TypeBottle
ConcentrationSlurry: 25%
MatrixMagnetic Agarose
Quantity100 mL
Shipping ConditionWet Ice
Binding CapacityUp to 80 mg of 6xHis-tagged Protein/mL of Settled Magnetic Beads
Particle Size30 μm
TypeMagnetic Affinity Bead
pH Range2 to 14 pH stability
Unit SizeEach
Contents & Storage
100 mL of 25% suspension. Store at 2–8°C.
Frequently asked questions (FAQs)
Can I automate purifications using Pierce High Capacity Ni-IMAC MagBeads, EDTA compatible?
The KingFisher product line has proven to give high-throughput, high purity, and consistency across samples when performing automated purifications using Pierce High Capacity Ni-IMAC MagBeads, EDTA compatible.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
With Pierce High Capacity Ni-IMAC MagBeads, EDTA compatible, which downstream assays are compatible with the eluted His-tagged protein?
Recovered protein can be run on SDS-PAGE and stained or analyzed by western blotting. Since the elution buffer contains 300 mM NaCl and 500 mM imidazole, recovered protein may need to be desalted or buffer-exchanged before proceeding to activity assays, mass spectrometry, or structural analyses.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
After regeneration of Pierce High Capacity Ni-IMAC MagBeads, EDTA compatible, is there a specific buffer in which the beads should be stored?
The recommended storage buffer for the magnetic beads is 20% ethanol, 10 mM sodium acetate, pH 6.50. Please refer to the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0024861_PierceHighCapNiIMACBeads_UG.pdf) for details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
With Pierce High Capacity Ni-IMAC MagBeads, EDTA compatible, how do I regenerate the beads?
Pierce High Capacity Ni-IMAC MagBeads, EDTA compatible are alkali-stable. The beads can be exposed to NaOH, washed with water, neutralized with PBS, washed with water, and put back into storage buffer. Please refer to the product manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0024861_PierceHighCapNiIMACBeads_UG.pdf) for details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How many uses/regenerations are possible with Pierce High Capacity Ni-IMAC MagBeads, EDTA compatible?
Up to 5 uses are possible with Pierce High Capacity Ni-IMAC MagBeads, EDTA compatible.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.