Genexus™ Dx Conical Bottles
Ion Torrent™
Genexus™ Dx Conical Bottles
This set of five conical bottles are installed in the sequencing reagents bay of the Genexus Dx Integrated Sequencer and serve as reservoirs for nucleotide reagent dilutions.
Have Questions?
Catalog NumberQuantity
A508691 Kit of 5
Catalog number A50869
Price (INR)
93,362.00
Each
Quantity:
1 Kit of 5
Price (INR)
93,362.00
Each
Genexus Dx Conical Bottles are installed in the sequencing reagents bay of the Genexus Dx Integrated Sequencer and serve as reservoirs for nucleotide reagent dilutions. They need to be replaced when filters in the bottles become partially clogged, causing decreased flow rate. If the sequencer detects restricted flow from the bottles during an instrument cleaning procedure, an alert will be displayed to replace the conical bottles.
For more information on when and how to replace the bottles, see the appendix sections of the Genexus Dx Integrated Sequencer User Guide for Dx or Assay Development software.
The Genexus Dx Integrated Sequencer is an in vitro diagnostic (IVD) medical device intended to be used by trained personnel in a professional laboratory environment for automated, qualitative targeted sequencing of human DNA or RNA extracted from formalin-fixed, paraffin-embedded (FFPE) tumor tissue samples or plasma samples.
For In Vitro Diagnostic Use. Not available in all countries or regions.
Specifications
Product TypeConical Bottles
Quantity1 Kit of 5
Sequencing TypeNext Generation Sequencing
Unit SizeEach