Attune™ CytPix™ Violet 6 Conversion Kit, for blue laser
Invitrogen™
Attune™ CytPix™ Violet 6 Conversion Kit, for blue laser
The Attune CytPix Violet 6 Conversion Kit is designed to reconfigure Attune NxT four-channel violet instruments to allow detection ofRead more
Have Questions?
Catalog Number
Quantity
A50926
1 Kit
Catalog number A50926
Price (INR)
580,209.00
Each
Add to cart
Quantity:
1 Kit
Price (INR)
580,209.00
Each
Add to cart
The Attune CytPix Violet 6 Conversion Kit is designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out by a Field Service Engineer.
Specifications
For Use With (Application)Flow Cytometry
For Use With (Equipment)Attune CytPix Flow Cytometer