Ion AmpliSeq™ SARS-CoV-2 Insight Research Assay - GS Manual
Ion Torrent™

Ion AmpliSeq™ SARS-CoV-2 Insight Research Assay - GS Manual

The Ion AmpliSeq SARS CoV 2 Insight Research Assay - GS Manual is a quick, accurate, and comprehensive next-generation sequencingRead more
Have Questions?
Catalog NumberQuantity
A5130596 reactions
Catalog number A51305
Price (INR)
1,713,201.00
Each
Quantity:
96 reactions
Price (INR)
1,713,201.00
Each
The Ion AmpliSeq SARS CoV 2 Insight Research Assay - GS Manual is a quick, accurate, and comprehensive next-generation sequencing (NGS) assay to enable research and surveillance of the SARS-CoV-2 virus, including variants of concern, through complete genome sequencing. The intelligent panel design includes new primers that broaden the coverage for variants of concern and increase the sensitivity of the panel for improved detection from lower viral titer samples.

This version of the assay is for manual library preparation. For automated library preparation using the Ion Chef System, please see Cat. No. A51306.

Features of the Ion AmpliSeq SARS CoV 2 Insight Research Assay - GS Manual include:
• Helps broaden NGS epidemiological surveillance to cover emerging SARS-CoV-2 variants through panel design improvements
• Improved limit of detection from various sample types due to increased sensitivity
• Enables greater access to NGS to rapidly obtain epidemiological insights with rapid turn-around time, workflow automation, and seamless informatics, lineage identification and data uploading to public SARS-CoV-2 data repositories for viral surveillance

With the emergence of the SARS-CoV-2 variants of concern (B.1.1.7, B.1.351, P.1) that may foreshadow increased transmissibility, disease severity, or impact on diagnostics, treatments, and vaccines, there is a critical need to sequence more SARS-CoV-2 samples. Researchers and public health agencies re leveraging Ion Torrent sequencing technology to discover, detect, and track emerging SARS-CoV-2 variants. Scientists must rapidly gain insights around the SARS-CoV-2 mutation rate, host infection risk, and viral spread in order to help reduce further impact of the virus.

The Ion AmpliSeq SARS-CoV-2 Insight Research Assay - GS Manual is part of an automated sample-to-variant report NGS workflow and provides >99% coverage of the viral genome and variants, including all serotypes. This panel consists of two pools of Ion AmpliSeq oligonucleotide primers specific to the SARS-CoV-2, five human expression controls, and one Ion AmpliSeq Library Kit Plus kit. The addition of new variant-tolerant primers enable this panel to be more robust for future variant tolerance over time.

Sufficient reagents are provided for the rapid preparation of 96 sample libraries. The panel also includes a plug-in suite complete with intuitive analysis tools for generating consensus sequence through multi-FASTA files and assigning variant lineage, helping make SARS-CoV-2 sequencing and epidemiology research accessible to labs regardless of NGS and informatics expertise.

The Ion AmpliSeq SARS CoV 2 Insight Research Assay - GS Manual is compatible with Torrent Suite Software 5.12 and later.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Ion AmpliSeq SARS‐CoV‐2 Insight Research Panel - GS Manual. Store in Freezer (-5°C to -30°C).
No. of Reactions96
Product LineIon AmpliSeq
Quantity96 reactions
SoftwareTorrent Suite Software 5.12
Unit SizeEach
Contents & Storage
• Ion AmpliSeq SARS‐CoV‐2 Insight Research Panel - GS Manual
• Ion AmpliSeq Library Kit Plus (96 reactions)

Store in freezer -5 to -30°C.