CTS™ AAV-MAX Lysis Buffer
Gibco™

CTS™ AAV-MAX Lysis Buffer

Gibco CTS AAV-MAX Lysis Buffer is a ready-to-use cell lysis reagent for the extraction of adeno-associated virus (AAV) particles fromRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A515210310 L
A5152101100 mL
A51521021 L
Catalog number A5152103
Price (INR)
1,035,537.00
Online offer
Ends: 30-Sep-2026
1,882,793.00
Save 847,256.00 (45%)
Each
Quantity:
10 L
Price (INR)
1,035,537.00
Online offer
Ends: 30-Sep-2026
1,882,793.00
Save 847,256.00 (45%)
Each

Gibco CTS AAV-MAX Lysis Buffer is a ready-to-use cell lysis reagent for the extraction of adeno-associated virus (AAV) particles from producer HEK293 cells. The cell lysis is detergent-based, eliminating the need for mechanical cell disruption, providing a milder and easier alternative when isolating AAV particles from cell cultures. The buffer is supplied as a 10X solution that can be directly added to HEK293 AAV production cultures to induce cell lysis.

Note: the formulation of this product differs from the formulation of our research-use-only AAV-MAX Lysis Buffer (Cat. No. A50520). This formulation reduces sample turbidity and precipitation formation post-clarification, including when samples are stored between clarification and subsequent downstream workflow steps.

Features of CTS AAV-MAX Lysis Buffer include:
• Polysorbate 20-based formulation that does not contain Triton X-100
• Formulated with 20 mM MgCl2 for convenience
• Manufactured in conformity with cGMP for medical devices (21 CFR Part 820) and follows USP and European Pharmacopoeia (Ph. Eur.) 5.2.12 recommendations. Drug Master File Letter of Authorization and Regulatory Support File available upon request.

The CTS AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production with research-grade and CTS versions to enable seamless transition from research to commercial manufacturing. Other components of the CTS AAV-MAX system include CTS Viral Production Cells 2.0, CTS AAV-MAX Transfection Kit, CTS Viral-Plex Complexation Buffer, and Viral Production Medium, AGT.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Concentration10X
Quantity10 L
Reagent TypeCell Lysis Buffer
Shelf Life12 Months
FormLiquid
Product LineAAV-MAX
Product TypeCell Lysis Buffer
pH6–8
Unit SizeEach
Contents & Storage
Store at 2–8°C.