Pierce™ FlexMix™ Calibration Solution for Auto-Ready Mass Spectrometers
Thermo Scientific™

Pierce™ FlexMix™ Calibration Solution for Auto-Ready Mass Spectrometers

The Thermo Scientific Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers is a room-temperature stable, ready-to-use liquid formulation for theRead more
Have Questions?
Catalog NumberQuantity
A517392 x 4 mL
Catalog number A51739
Price (INR)
34,379.00
Each
Quantity:
2 x 4 mL
Price (INR)
34,379.00
Each
The Thermo Scientific Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers is a room-temperature stable, ready-to-use liquid formulation for the calibration of Thermo Scientific mass spectrometers with the integrated Auto-Ready ion source.

Pierce FlexMix Calibration Solution features include:
Improved mass accuracy—extended mass range improves mass accuracy
Positive and negative mode calibration—single formulation with extended mass ranges for both ionization calibration modes
Highly validated—high-quality, rigorously-tested formulation with lot-specific certificates of analysis
Ready-to-use—bottle may be directly loaded into the instrument for scheduled automatic calibration
Stable—room temperature for 18 months and within the Auto-Ready mass spectrometer for up to 4 months

The Pierce FlexMix Calibration Solution is a ready-to-use formulation that has been optimized for both positive and negative ionization calibration of instruments. The extended range masses (50 to 3000 m/z) improve the sensitivity and mass accuracy of Orbitrap mass spectrometers. The solution is now available in an Auto-Ready format for the Orbitrap IQ-X Mass Spectrometer using Orbitrap Tribrid Series 3.5 Instrument Control Software.

For hardware-related support visit: www.unitylabservices.com.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorColorless
Physical FormLiquid
Quantity2 x 4 mL
Percent PurityMixture
Unit SizeEach

Frequently asked questions (FAQs)

In which mass spectrometer instruments can I use the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers?

Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers is designed to be used in mass spectrometer instruments with autocalibration feature, such as Orbitrap IQ-X Mass Spectrometer using Orbitrap Tribrid Series 3.5 Instrument Control Software (or later).

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

Does the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers need to be diluted?

Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers is a ready to use reagent and there is no need for dilution.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

Is there a specific mode in which Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers needs to be run?

Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers can be run in Positive or Negative mode.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

What is the difference between Pierce FlexMix Calibration Solution and Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers?

Both the Pierce FlexMix Calibration Solution and Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers have the same 16 components and produce the same spectra. However, the Pierce FlexMix Calibration Solution for Auto-Ready Mass Spectrometers has a different storage solution: LC-MS grade acetonitrilie 87% (v/v) and LC-MS grade water 13% (v/v). The FlexMix Calibration for Auto-Ready Mass Spectrometers is designed to be used only in mass spectrometer instruments with autocalibration feature, such as Orbitrap IQ-X Mass Spectrometer using Orbitrap Tribrid Series 3.5 Instrument Control Software (or later).

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.