MagMAX™ Dx Viral/Pathogen NA Isolation Kit
Applied Biosystems™

MagMAX™ Dx Viral/Pathogen NA Isolation Kit

The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is specifically designed to recover viral and bacterial nucleic acids fromRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A52735Up to 2,000 preps
A52736Up to 600 preps
Catalog number A52735
Price (INR)
334,077.00
Each
No. of Reactions:
Up to 2,000 preps
Price (INR)
334,077.00
Each
The Applied Biosystems MagMAX Dx Viral/Pathogen NA Isolation Kit is specifically designed to recover viral and bacterial nucleic acids from human respiratory biological specimens for qPCR downstream applications. The kit uses MagMAX magnetic-bead technology to enable reproducible recovery of high-quality nucleic acids.

Features of the MagMAX Dx Viral/Pathogen NA Isolation Kit include:
• For in vitro diagnostic use. This product is US IVD compliant
• For isolation and purification of viral and bacterial nucleic acids from a range of respiratory biological specimens such as nasopharyngeal swab, anterior nasal swab, sputum, and saliva
• Single, flexible automation protocol that accommodates sample volume inputs of 200–400 μL
• Fast, automated procedures that allow 96 samples to be processed on the KingFisher Apex Dx Purification Instrument with ∼30-minute instrument run time

Access the Instructions for Use
(You will be redirected to sign into the resource library.)
Access the MagMAX Dx protocol for KingFisher Apex Dx instrument: Dx_MVP_RSP_APX_V1.kfdx
(You will be redirected to sign into the resource library.)
More information about the KingFisher Apex Dx Purification Instrument

Please check with your country specific representative for more information on product registration.

For In Vitro Diagnostic Use
Specifications
For Use With (Application)Real-Time PCR
For Use With (Equipment)KingFisher™ Apex Dx
No. of ReactionsUp to 2,000 preps
Nucleic Acid TypeViral and Bacterial
Product LineMagMAX
QuantityUp to 2,000 preps
Sample TypeNasopharyngeal swab, Anterior nasal swab, Saliva, Sputum
Shelf Life24 Months
Shipping ConditionRoom Temperature
TargetRespiratory Pathogens
Format96-well Plate
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage

• Binding Solution, 530 mL
• Wash Solution, 1000 mL
• Binding Beads, 20 mL
• Proteinase K, 10 mL
• Elution Buffer, 120 mL

Shipped and stored at room temperature.