RapidHIT™ ID Primary Cartridge GlobalFiler™ Express 150 Kit
Applied Biosystems™
RapidHIT™ ID Primary Cartridge GlobalFiler™ Express 150 Kit
The Applied Biosystems™ RapidHIT™ ID Primary Cartridge GlobalFiler Express 150 Kit is used with the Applied Biosystems RapidHIT ID SystemRead more
Have Questions?
Catalog NumberQuantity
A53084150 runs
Catalog number A53084
Price (INR)
1,279,713.00
Each
Quantity:
150 runs
Price (INR)
1,279,713.00
Each
The Applied Biosystems™ RapidHIT™ ID Primary Cartridge GlobalFiler Express 150 Kit is used with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to generate lab-quality forensic DNA profiles from reference samples using the ACE GlobalFiler Express Sample Cartridge (Cat. No. A41831) and from crime scene samples using the RapidINTEL™ Sample Cartridge (Cat. No. A43942).     
Each GlobalFiler™ Express 150 Primary Cartridge Kit contains reagents for use with 150 sample cartridges.  Once installed in the Applied Biosystems RapidHIT ID System, this primary cartridge remains in place for 150 runs before needing to be replaced.      
Easy to assemble and install
Automatically tracks number of runs, lot number, and shelf life
Allows user to run 150 runs with no further action after installation
For Forensic and Human Identity Applications Only.
Specifications
For Use With (Application)DNA Fragment Analysis
For Use With (Equipment)RapidHIT ID System
Product LineRapidHIT
Product TypePrimary cartridge
Quantity150 runs
Unit SizeEach
Contents & Storage
  • 1 Primary Cartridge Housing – GlobalFiler Express 150 (room temperature)
  • 2 Utility Cartridges (room temperature)
  • 1 Gel Cartridge (refrigerated)
  • 1 ACE GlobalFiler Express Control Cartridge (refrigerated)
  • 1 Positive Control RapidHIT ID ACE GlobalFiler Express Sample Cartridge (refrigerated)