AccelerOme™ System Test Kit
AccelerOme™ System Test Kit
AccelerOme™ System Test Kit
Thermo Scientific™
AccelerOme™ System Test Kit
The Thermo Scientific AccelerOme System Test Kit provides control reagents for installation and QC of the AccelerOme Automated Sample PrepRead more
Have Questions?
Catalog NumberQuantity
A532201 Kit
Catalog number A53220
Price (INR)
15,830.00
Each
Quantity:
1 Kit
Price (INR)
15,830.00
Each
The Thermo Scientific AccelerOme System Test Kit provides control reagents for installation and QC of the AccelerOme Automated Sample Prep System.
Features of the Accelerome System Test Kit include:
• Convenient—ready-to-use solutions provided in a tray that is directly inserted into the instrument
• Predictive—buffer matrix and peptide quantitation calibration standard monitor liquid handling and UV spectrophotometer system performance
• Integrated—designed specifically to provide installation and QC support of the AccelerOme system
• Stable—provided in stable liquid format
The AccelerOme System Test Kit is required for installation of the AccelerOme Automated Sample Prep System. The kit contains a matrix buffer and peptide quantitation standard to assess the system’s liquid handling reproducibility and accuracy. Additionally, the quantitation standard serves as a quality control standard for the on-board UV spectrophotometer. The system test kit may also be used for troubleshooting and routine instrument calibration.
The AccelerOme integrated system provides robust proteomic mass spec sample preparation. The instrument hardware is built on a proven liquid handling format for reproducible sample processing and optimized on-line μSPE sample clean-up for excellent peptide recovery. Peptide quantitation post-clean-up is achieved through accurate UV spectrophotometric analysis to determine peptide concentrations prior to LC-MS analysis. The complete system (kits + reagents + software) was designed to provide high quality peptides while improving productivity with an easy-to-use automated system.
Specifications
Quantity1 Kit
Detection MethodUV Absorption
Product LineThermo Scientific
Product TypeReagent Kit
Starting MaterialProtein Samples
Unit SizeEach
Frequently asked questions (FAQs)
Can the AccelerOme System Test Kit be used on a different platform such as Agilent?
The AccelerOme System Test Kit has been optimized exclusively for the AccelerOme Automated Sample Prep Platform.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
How often do I need to use the AccelerOme System Test Kit?
The AccelerOme System Test Kit is meant to be used for calibration (by FAS/FSE) when the AccelerOme Automated Sample Prep System is installed or restarted after maintenance.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Can I reuse the AccelerOme System Test Kit?
We do not recommend reusing the AccelerOme System Test Kit. It is meant to be used for calibration (by FAS/FSE) when the AccelerOme Automated Sample Prep System is installed or restarted after maintenance.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
In which format is the AccelerOme System Test Kit provided?
The AccelerOme System Test Kit is a ready-to-use tray to be inserted in the AccelerOme Automated Sample Prep System, to start the calibration.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.