Product Image
Applied Biosystems™
GeneMapper™ Software v6.1, client installation
Note: This client version of GeneMapper Software v6.1 installs the client configuration (user interface) only. Users must install GeneMapper SoftwareRead more
Have Questions?
Catalog NumberQuantity
A535501 license
Catalog number A53550
Price (INR)
442,926.00
Each
Quantity:
1 license
Price (INR)
442,926.00
Each
Note: This client version of GeneMapper Software v6.1 installs the client configuration (user interface) only. Users must install GeneMapper Software v6.1, full installation, prior to installing the client version, which should be installed on a computer separate from that of the full version.
GeneMapper Software is a flexible genotyping software package that provides DNA sizing and quality allele calls for all of our electrophoresis-based genotyping systems. This software specializes in multi-application functionality, including amplified fragment length polymorphism (AFLP), loss of heterozygosity (LOH), microsatellite and SNP genotyping analysis. GeneMapper Software can help users increase data processing efficiency with remote auto-analysis and command line operation, and allows for multiuser, client-server deployment. The software uses Process Quality Values (PQVs) for automated identification that reduces data review time for high throughput genotyping. In addition, the security and audit features help users meet 21 CFR 11 requirements.
Features include:
• Installation on the data collection computer or a non-instrument computer
• Support of networked multiple clients when installed on a non-instrument computer
• An Oracle™ database with project-related data storage that ensures faster installation times and flexible archive capabilities
• Customizable project auto-saving frequency
• Defining of linearity range in the analysis methods for more reproducible, higher quality sizing of larger fragments (>800bps)
• Analysis of up to 8-dye data
GeneMapper Software has not undergone specific developmental validation for human identification applications. Human identification laboratories analyzing single-source or parentage samples that choose to use GeneMapper Software for data analysis should perform their own developmental validation studies.
Several users may access the database concurrently; however, because genotype analysis is data-intensive, more than five clients simultaneously accessing the database may slow the performance of GeneMapper Software.
The software is tested with the following specifications:
• Dell OptiPlex XE3 Windows™ 10 Enterprise LTSC operating system
• Dell 3551 Windows 10 Enterprise LTSC operating system
• CPU: Core i5-6440HQ/3.0 GHz and CoreTM i5-10400H CPU @ 2.60 GHz
• Memory: 16 GB
• Disk: Free space: 500 MB on the boot drive, 7 GB on the drive on which the GeneMapper Software is installed
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Data EntryCE Fragment Analysis (.fsa files)
LicenseClient
Operating SystemWindows 10
Product TypeGeneMapper Software 5 Client Installation
Quantity1 license
Software CategorySupports files from all AB Genetic Analyzers
Product LineGeneMapper
Software TypeCE Fragment Analysis
Unit SizeEach
Contents & Storage
The software is provided in the form of a USB stick.
Frequently asked questions (FAQs)
How can I set up auto-analysis for data in GeneMapper with the 3500 Data Collection Software version 3.x?
Beginning with 3500 Data Collection Software version 3, the 3500 Data Collection Software does not support automatic secondary analysis. This is also indicated in the software release notes. To perform secondary data analysis, you will need to manually import the .fsa file into the GeneMapper software. For optimal performance, we recommend that you install the GeneMapper Software on a stand-alone computer that is not part of the 3500 Genetic Analyzer system.
Find additional tips, troubleshooting help, and resources within our Fragment Analysis Software Support Center.
Can I install GeneMapper Software v6.1, full installation and GeneMapper Software v6.1, client installation on the same computer?
No, GeneMapper Full Software and GeneMapper Client Software must be installed on different computers.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Where can I download the Release Notes for GeneMapper Software v6.1, client installation?
Please download this PDF (http://downloads.thermofisher.com/Instrument_Software/Genemapper_6.1_Client/100107867_GeneMapper-Client_v6.1_Release_Notes.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
We just upgraded to GeneMapper Software v6.1, and I'm wondering if my old data files will work on it.
GeneMapper Software v6.1 is compatible with the following files:
- 3130 series, Data Collection Software v3.0, 3.1, 4
- 3730 series, Data Collection Software v3.x, 4.x
- 3500 series, Data Collection Software v1.x, 2, 3.x
- SeqStudio Data Collection Software v1.x
- SeqStudio Flex Series Genetic Analyzer, Data Collection Software v1.x
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Can I install GeneMapper Software v6.1 on Windows 7 operating system?
No, GeneMapper Software v6.1 supports only Windows 10 Professional/Enterprise operating system.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.