SeqStudio™ 8 Flex Genetic Analyzer, desktop, SmartStart Orientation, 1-year AB Assurance with Planned Maintenance
Applied Biosystems™
SeqStudio™ 8 Flex Genetic Analyzer, desktop, SmartStart Orientation, 1-year AB Assurance with Planned Maintenance
The Applied Biosystems SeqStudio 8 Flex Genetic Analyzer offers design improvements and technological advances to the capillary electrophoresis (CE) workflow.Read more
Have Questions?
Catalog NumberQuantity
A537901 package
Catalog number A53790
Price (INR)
-
Quantity:
1 package
The Applied Biosystems SeqStudio 8 Flex Genetic Analyzer offers design improvements and technological advances to the capillary electrophoresis (CE) workflow. It delivers gold-standard Sanger sequencing and fragment analysis data quality with increased flexibility, easier operation, and cloud connectivity to enable researchers to manage their workloads and work/life balance effectively. Designed for ultimate ease-of-use, this mid-throughput system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab.
This system package includes a one-year AB Assurance extended warranty with Planned Maintenance and Smart Start orientation.
Features of the SeqStudio 8 Flex Genetic Analyzer include:
• Space savings with true standalone operation; setup, start, monitor, and analyze directly on the touchscreen
• Improve workload management with continuous plate loading and sample reprioritization
• Realize lab efficiencies with remote monitoring
• New, improved capillary arrays for simple and hassle-free handling
• Reduce downtime with integrated remote troubleshooting tools
Easy-to-use data collection software
The data collection software pre-installed on the on-board touch screen computer takes user-friendliness to a new level. It offers an intuitive dashboard design, simplified plate set-up, and embedded self-help videos for common operations so that new users can get up and running quickly.
Scheduling flexibility
Four-plate capacity, continuous plate loading, and urgent sample reprioritization provide both the largest mid-throughput capacity and complete scheduling flexibility, translating into extra time in your day.
Remote functionality
Our cloud-based Connect platform provides enhanced flexibility by allowing users to set up plate files, monitor instrument and consumables performance and run progress, and even perform some critical instrument functions from anywhere.
Stay connected to your data
The SeqStudio 8 Flex system offers an automated workflow to obtain and share results from existing secondary analysis applications on Connect. These web-based software applications include Peak Scanner, Microsatellite Analysis, NGS Confirmation, Quality Check and Variant Analysis. They are compatible with Mac and PC.
Peace of mind with Smart service
Minimize downtime with premium service offerings for the SeqStudio 8 Flex Genetic Analyzer. Smart Help allows users to contact technical support directly from the touchscreen, without the need to download files or spend time on the phone. Experienced technicians will analyze the request and respond according to the user’s preference (phone or email) and resolve the issue as quickly as possible.
Smart Remote Support is an immersive collaboration tool that allows customers to show issues quickly and efficiently to technical support for resolution, helping reduce instrument downtime and minimize the need for on-site visits.
High application versatility
Simple workflows, reliable assays and reagents, and intuitive software are available for a broad range of applications, including multiplexed PCR analysis, next-generation confirmation, species identification, STR genotyping, cell-line authentication, dsDNA quality control, plasmid sequencing, and genome editing analysis.
Upgrade capabilities when you need them
The SeqStudio 8 Flex Genetic Analyzer can be seamlessly upgraded to a SeqStudio 24 Flex Genetic Analyzer by a field service engineer to provide additional throughput as your workload increases.
Results you can trust
Well-to-well and instrument-to-instrument data accuracy is achieved with the proven Applied Biosystems capillary electrophoresis technology.
Service plans help protect your investment
• Maximize system uptime with fast repair turnaround time from manufacturer-trained and certified field service engineers (FSEs)
• Extend the life of your instrument and help keep it running at peak performance with annual maintenance service
During the annual maintenance, the FSE will update the software, if required; clean, lubricate, and align relevant parts and optical systems; and perform functional tests while documenting tasks, results, and findings.
Specifications
FilteredFiltered
Quantity1 package
SterilityNon-sterile
Total Service Coverage1 year manufacturer warranty and 1 year of field service with PM
Voltage100/240 V
Unit SizeEach
Frequently asked questions (FAQs)
How can I request assistance from Technical Support or Instrument Service for SeqStudio Flex Series Genetic Analyzers?
You can request assistance from Technical Support or Instrument Service on the SeqStudio Flex Series Genetic Analyzers via the Smart Help function. In order to use this function, access to Thermo Fisher Connect must be enabled. You can also contact Technical Support at techsupport@thermofisher.com or Service Support at instrumentsupport@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our SeqStudio Flex Series Genetic Analyzers Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
With the SeqStudio Flex Genetic Analyzer, I tried to start a run, but the instrument displays a notification that there is not enough storage space. What should I do?
The SeqStudio Flex Genetic Analyzer instrument will not run unless the required amount of space is available to save the results on the instrument. To free up space on the hard drive, you will need to delete the run histories. We recommend that you first export the results before deleting them as the process is irreversible. At the home screen, tap Actions > Run history. Select the plates that you want to delete. Tap Delete > OK to confirm.
Find additional tips, troubleshooting help, and resources within our SeqStudio Flex Series Genetic Analyzers Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
In the SeqStudio Flex Genetic Analyzer, I do not have access to the CBC or capillary array because the CBC is currently beneath the array with the tips inserted. How can I move it so I can have access to the array or CBC?
You can move the CBC by using the Manual commands. Tap on Dashboard > Instrument Status > Manual commands. Select Move buffer container > Send command. Once the CBC is no longer beneath the array with the tips inserted, close the Manual commands screen. You will now have access to the array and CBC.
Find additional tips, troubleshooting help, and resources within our SeqStudio Flex Series Genetic Analyzers Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
What should I do if the SeqStudio Flex Genetic Analyzer touchscreen becomes unresponsive?
If the SeqStudio Flex Genetic Analyzer touchscreen becomes unresponsive, press the power on/off button on the front of the instrument to restart the instrument. If the touchscreen is still unresponsive after the reboot, contact Technical Support at techsupport@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our SeqStudio Flex Series Genetic Analyzers Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
What should I do if the SeqStudio Flex Genetic Analyzer instrument panel is blinking amber?
If the SeqStudio Flex Genetic Analyzer instrument panel is blinking amber, you should restart the instrument.
Find additional tips, troubleshooting help, and resources within our SeqStudio Flex Series Genetic Analyzers Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.