Ion SingleSeq™ 96 Kit LUN
Product Image
Ion Torrent™

Ion SingleSeq™ 96 Kit LUN

The Ion SingleSeq 96 Kit LUN is designed for low-cell-number input library construction prior to low-pass genome sequencing detection ofRead more
Have Questions?
Catalog NumberQuantity
A5427196 samples
Catalog number A54271
Price (INR)
-
Quantity:
96 samples
The Ion SingleSeq 96 Kit LUN is designed for low-cell-number input library construction prior to low-pass genome sequencing detection of copy number events. Requiring as little as 6 pg of DNA from a single cell, or input from multiple cells, this kit includes reagents to extract, amplify, and barcode genomic DNA for preparation of libraries from up to 96 samples.

The Ion SingleSeq 96 Kit LUN is included with the ReproSeq PGS Kit for the detection of chromosomal aneuploidies and chromosome arm copy number events (>48 Mb). The ReproSeq PGS Kit is used with the Ion Chef System and the Ion S5 sequencing system to enable automated template preparation, chip loading, and sequencing with disposable reagent cartridges for fast and simple instrument set-up to minimize hands-on time.

Compatible kits include:
• ReproSeq PGS Kit with Ion 520 Chips GPR (24 samples/run) (Cat. No. A53172)
• ReproSeq PGS Kit with Ion 530 Chips GPR (96 samples/run) (Cat. No. A53173)

For Laboratory Use
Specifications
For Use With (Equipment)Ion GeneStudio S5 System, Ion Chef™ System
LibrariesFragment Library
No. of Reactions96
Product LineReproSeq
Product Type96 Kit
Quantity96 samples
Shipping ConditionApproved for shipment on Wet or Dry Ice
Throughput96 Samples
Workflow StepLibrary Generation
Sequencing TypeGenome and DNA Sequencing
Unit SizeEach
Contents & Storage
Store at –10 to –30°C.