RapidINTEL™ Plus Negative Control Cartridge
Applied Biosystems™

RapidINTEL™ Plus Negative Control Cartridge

The Applied Biosystems RapidINTEL Plus Negative Control Cartridge is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to verify instrument and/or reagent performance.
Have Questions?
Catalog NumberQuantity
A543431 cartridge
Catalog number A54343
Price (INR)
28,102.00
Each
Quantity:
1 cartridge
Price (INR)
28,102.00
Each

The Applied Biosystems RapidINTEL Plus Negative Control Cartridge is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to verify instrument and/or reagent performance. Each cartridge is individually wrapped and single use/disposable.

Required for use
• Applied Biosystems RapidHIT ID System (Cat. No. A41810)
• RapidHIT ID Instrument Software v2.0 or higher
• RapidHIT ID Primary Cartridge (Cat. No. A53084)

For Research Use Only.
Specifications
For Use With (Application)Human Identification
For Use With (Equipment)RapidHIT™ ID
Quantity1 cartridge
Shelf Life12 Months
FormatCartridge
Unit SizeEach