AAV-MAX Helper-Free AAV Production System Kit, without lysis buffer
Gibco™

AAV-MAX Helper-Free AAV Production System Kit, without lysis buffer

The Gibco AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production. It is based onRead more
Have Questions?
Catalog NumberQuantity
A548241 Kit
Catalog number A54824
Price (INR)
150,955.00
Online offer
Ends: 31-Dec-2026
201,273.00
Save 50,318.00 (25%)
Each
Quantity:
1 Kit
Price (INR)
150,955.00
Online offer
Ends: 31-Dec-2026
201,273.00
Save 50,318.00 (25%)
Each
The Gibco AAV-MAX Helper-Free AAV Production System is a scalable, high-titer platform for suspension AAV production. It is based on a high-density suspension culture of clonal 293F-derived Viral Production Cells 2.0 in Viral Production Medium. Optimal viral titers are mediated by our advanced lipid nanoparticle transfection reagent in combination with a novel production enhancer. The components are designed to work synergistically, resulting in maximal titers and eliminating the need to optimize reagents and protocols.

Features of the AAV-MAX Helper-Free AAV Production System include:
• Components work synergistically to produce high AAV titers greater than 5 x 1010 vg/mL (unpurified)
• Suspension-based system has scalable protocols from shake flask to bioreactor scale
• Simplified workflow—streamlined protocol using helper virus-free triple transfection
• Clonal, 293F-derived, high-producer cell line
• Robust transfection protocols are adaptable to large-scale transfection

The components included in the AAV-MAX System Kit are:
• 1 vial of Viral Production Cells 2.0
• 1 L of Viral Production Medium
• 1 AAV-MAX Transfection Kit for 1 L of production
• 100 mL Viral-Plex Complexation Buffer
• 100 mL GlutaMAX Supplement

All components are also available for purchase separately. Note, this version of the kit does not contain the AAV-MAX Lysis Buffer. To purchase the kit including the AAV-MAX Lysis Buffer, please see Cat. No. A51217.


Specifications
Cell LineViral Production Cells 2.0
Cell TypeKidney (Embryonic)
For Use With (Application)AAV production
Product LineAAV-MAX
Product TypeAAV Production System
Quantity1 Kit
Shelf Life12 months from date of manufacture
Culture TypeSuspension Cell Culture
SterilitySterile-filtered
Unit SizeEach
Contents & Storage
• 1 vial Viral Production Cells 2.0
• 1 L Viral Production Medium
• 1 AAV-MAX Transfection kit (for 1 L production)
• 100 mL Viral-Plex Complexation Buffer
• 100 mL GlutaMAX