RapidHIT™ ID Booking Station ACE GlobalFiler™ Express Complete 150-Sample Kit
Applied Biosystems™

RapidHIT™ ID Booking Station ACE GlobalFiler™ Express Complete 150-Sample Kit

The Applied Biosystems RapidHIT ID Booking Station ACE GlobalFiler Express Complete 150-Sample Kit is for use with the Applied BiosystemsRead more
Have Questions?
Catalog NumberQuantity
A570751 Kit
Catalog number A57075
Price (INR)
-
Quantity:
1 Kit

The Applied Biosystems RapidHIT ID Booking Station ACE GlobalFiler Express Complete 150-Sample Kit is for use with the Applied Biosystems RapidHIT ID System (Cat. No. A41810) to generate lab-quality forensic DNA profiles from arrestee samples collected during the booking process. Each kit contains all the cartridges necessary to run 150 arrestee samples.

Features of the Applied Biosystems RapidHIT ID Booking Station ACE GlobalFiler Express Complete 150-Sample Kit include:
• Develop DNA profiles from arrestees in the booking station in just 90 minutes
• Upload arrestee DNA profiles to CODIS directly from the booking station
• Link arrestees to previous crimes while still in custody

Requirements:
• Applied Biosystems RapidHIT ID System (Cat. No. A41810)
• Applied Biosystems RapidHIT ID DNA Booking Station instrument software v1.0 or higher

Specifications
For Use With (Application)Booking station arrestee testing
For Use With (Equipment)RapidHIT ID
No. of Samples150 Samples
Quantity1 Kit
Sample TypeBuccal Swabs
Shelf Life12 Months
FormatKit
Unit SizeEach
Contents & Storage
• 3 x ACE GlobalFiler Express 50-Sample Kit w/ Swab (refrigerated)
• Primary Cartridge Housing – GlobalFiler Express 150 (room temperature)
• 2 x Utility Cartridges (room temperature)
• Gel Cartridge (refrigerated)
• GlobalFiler Express Control (Ladder) Cartridge (refrigerated)