Pierce™ Dilution-Free™ Tubestrip Holder
Thermo Scientific™
Pierce™ Dilution-Free™ Tubestrip Holder
The Pierce Dilution-Free Tubestrip Holder is a reusable foam stand for use with the Pierce Dilution-Free BSA Protein Standards. ProteinRead more
Have Questions?
Catalog NumberQuantity
A571841 each
Catalog number A57184
Price (INR)
1,407.00
Each
Quantity:
1 each
Price (INR)
1,407.00
Each
The Pierce Dilution-Free Tubestrip Holder is a reusable foam stand for use with the Pierce Dilution-Free BSA Protein Standards. Protein standard concentrations are printed on the Tubestrip Holder for easy reference and pipetting on the benchtop.
Pierce Dilution-Free BSA Protein Standards
Pierce Dilution-Free BSA Protein Standards are high-quality bovine serum albumin (BSA) reference samples for generating accurate standard curves and calibration controls in total protein assays. They are packaged as a set of seven pre-diluted standards in a multichannel pipette-friendly tubestrip, eliminating the need for serial dilutions. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.
Related products
Pierce Dilution-Free BSA Protein Standards, Multichannel Pipette Compatible, 0.125–2 mg/mL
Pierce Dilution-Free BSA Protein Standards, Multichannel Pipette Compatible, 0.125–10 mg/mL
For Research Use Only.
Specifications
Product LinePierce Dilution-Free
Product TypeTubestrip holder
Quantity1 each
Unit SizeEach