SureTect™ Beverage Spoilage Multiplex PCR Assay
Thermo Scientific™
SureTect™ Beverage Spoilage Multiplex PCR Assay
Rapid qPCR solutions for beverage manufacturers. Rapidly detect a broad range of beverage spoilers from one PCR reaction with this assay.
Have Questions?
Catalog NumberQuantity
A57289-TF
also known as A57289
96 tests
Catalog number A57289-TF
also known as A57289
Price (INR)
-
Quantity:
96 tests
A rapid solution to rapidly detect and differentiate yeast and mold, Brettanomyces spp., preservative-resistant yeast, lactic acid bacteria, and acetic acid bacteria in aseptically produced and carbonated product samples. Developed in partnership with a major global beverage manufacturer to meet the industry’s needs.
The SureTect™ Beverage Spoilage Multiplex PCR Assay is an optimized solution combining speed and performance in an easy-to-use and cost-effective platform, giving robust results that you can always trust.
Simple
  • Easy-to-use, simple workflow
  • Pre-dispensed lysis reagents and lyophilized PCR reagents
  • Automatic lysis cycle with very few pipetting steps
Fast and flexible
  • Fast screening and differentiation of yeast and mold in a single PCR test
  • Rapid detection for early actions and high confidence in production
  • Test up to 96 samples in each run
  • Open PCR platforms
Smart
  • Highly specific multiplex qPCR to reduce cost and hands-on time, providing results from the four most important spoilage microorganism families for beverage, including over 100 strains, in one PCR reaction:
    • Yeast and molds (Y&M)
    • Lactic acid bacteria (LAB) and acetic acid bacteria (AAB)
    • Brettanomyces species
    • Preservative-resistant yeasts (PRY)
  • Actionable results up to 4 days earlier (depending on your current workflow) leading to risk-based decisions
  • Optimized workflow with pierceable seals to reduce cross-contamination
  • Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
  • Actionable data supports fast decisions in a complex product supply chain
  • Internal positive control (IPC) in every assay for complete confidence in results
The SureTect™ Beverage Spoilage Multiplex PCR Assay includes:
  • Lysis Plus Tubes
  • Lysis Supplement
  • Beverage Spoilage Multiplex PCR Assay Tubes
  • Thermo Scientific 96-well Pierceable Seals (3 sheets per kit)
  • SureTect PCR Caps
The SureTect™ Beverage Spoilage Multiplex PCR Assay is validated on the Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System with RapidFinder™ Analysis Software v3.0 or later, allowing you to test easily and confidently. After completing the run, interpreted results are clearly displayed and can be reported, stored, printed, or downloaded as required.
For Testing of Food and Environmental Samples Only
Specifications
Quantity96 tests
Unit SizeEach
Contents & Storage
Store the unopened kits at 2–8°C. Store the individual components as indicated in the instructions for use.