QuantStudio™ Absolute Q™ AutoRun Digital PCR Suite, single to dual configuration upgrade
Applied Biosystems™
QuantStudio™ Absolute Q™ AutoRun Digital PCR Suite, single to dual configuration upgrade
Upgrade your existing QuantStudio Absolute Q AutoRun Digital PCR Suite, Single Configuration to the QuantStudio Absolute Q AutoRun Digital PCR Suite, Dual Configuration, an integrated system solution that scales precise nucleic acid quantification and offers intuitive automation to help support productivity and accelerate progress.
Have Questions?
Catalog NumberQuantity
A576121 upgrade
Catalog number A57612
Price (INR)
2,871,794.00
Each
Quantity:
1 upgrade
Price (INR)
2,871,794.00
Each
Upgrade your existing QuantStudio Absolute Q AutoRun Digital PCR Suite, Single Configuration, to the QuantStudio Absolute Q AutoRun Digital PCR Suite, Dual Configuration, an integrated system solution that scales precise nucleic acid quantification and offers intuitive automation to help support productivity and accelerate progress.
The QuantStudio Absolute Q AutoRun Digital PCR Suite, Dual Configuration, consists of two QuantStudio Absolute Q Digital PCR systems integrated with the Spinnaker Microplate Mover via the easy-to-use Momentum Workflow Scheduler Software to manage loading and unloading of plates for an automated workflow. By harnessing the power of two digital PCR systems in your automated workflow, you can double your throughput while fortifying the resilience of your operations.
This upgrade includes a location bracket, a nest adapter, necessary cables, and an updated Thermo Scientific Momentum Workflow Software license to support integration of the second QuantStudio Absolute Q Digital PCR System.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Calibration TypePositional Teaching
For Use With (Application)Digital PCR
For Use With (Equipment)Absolute Q™
Quantity1 upgrade
Voltage100/240 V
Unit SizeEach