SeqStudio™ 24 Flex Genetic Analyzer for Pharma Analytics
Applied Biosystems™
SeqStudio™ 24 Flex Genetic Analyzer for Pharma Analytics
The Applied Biosystems SeqStudio 24 Flex Genetic Analyzer for Pharma Analytics offers design improvements and technological advances to the capillaryRead more
Have Questions?
Catalog NumberQuantity
A578301 each
Catalog number A57830
Price (INR)
24,420,616.00
Each
Quantity:
1 each
Price (INR)
24,420,616.00
Each
The Applied Biosystems SeqStudio 24 Flex Genetic Analyzer for Pharma Analytics offers design improvements and technological advances to the capillary electrophoresis (CE) workflow that can be used for pharma analytics. It delivers gold-standard Sanger sequencing and fragment analysis data quality with increased flexibility, easier operation, and cloud connectivity to enable researchers to manage their workloads and work/life balance effectively. Designed for ultimate ease-of-use, this mid-throughput system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab. The SeqStudio 24 Flex analyzer is compatible with MicroSEQ Microbial Identification Software for right-the-first-time microbial identification.
Features of the SeqStudio 24 Flex Genetic Analyzer for Pharma Analytics include:
• Space savings with true standalone operation
• Set up, start, monitor, and analyze directly on the touchscreen
• Improve workload management with continuous plate loading and sample reprioritization
• Realize lab efficiencies with remote monitoring
• Improved capillary arrays for simple and hassle-free handling
• Can reduce downtime with integrated remote troubleshooting tools
• SAE software—security, audit, and e-signature functionality for protection and traceability
This package includes:
• 24-capillary SeqStudio Flex Genetic Analyzer
• SAE Administrator Console Software v2.1, compatible with MicroSEQ ID Microbial Identification Software
Superb microbial identification using MicroSEQ microbial identification system
The SeqStudio Flex Genetic Analyzer is part of the MicroSEQ microbial identification system for the sequencing of bacterial and fungal organisms. This system uses a highly accurate phylogenetic approach for microbial identification based on the sequencing of the 16S rRNA gene for bacteria or the D2 region of the large subunit ribosomal DNA for fungi. The resulting sequences of microbial samples are compared to sequences in our curated and validated collection. The MicroSEQ ID microbial identification system allows you to easily identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library.
Easy-to-use data collection software
The data collection software pre-installed on the on-board touchscreen computer takes user-friendliness to a new level. It offers an intuitive dashboard design, simplified plate setup, and embedded self-help videos for common operations to enable new users can get up and running quickly.
Scheduling flexibility
Four-plate capacity, continuous plate loading, and urgent sample reprioritization can provide both a large mid-throughput capacity and complete scheduling flexibility, translating into extra time in your day.
Remote functionality
Our cloud-based Connect platform can provide enhanced flexibility by allowing users to set up plate files, monitor instrument and consumables performance and run progress, and even perform some critical instrument functions from anywhere.
Peace of mind with Smart service
Our premium service offerings for the SeqStudio 24 Flex Genetic Analyzer. Smart Help allows users to contact technical support directly from the touchscreen, without the need to download files or spend time on the phone, helping minimize downtime. Experienced technicians can analyze the request and respond according to the user’s preference (phone or email) and help resolve the issue as quickly as possible.
Smart Remote Support is an immersive collaboration tool that allows customers to show issues quickly and efficiently to technical support for resolution, helping reduce instrument downtime and minimize the need for on-site visits.
Results you can trust
Well-to-well and instrument-to-instrument data accuracy can be achieved with the proven Applied Biosystems capillary electrophoresis technology.
For Research Use Only.
Specifications
FilteredFiltered
Quantity1 each
SterilityNon-sterile
Voltage100/240 V
Unit SizeEach