TaqMan™ Cells-to-CT™ Express Lysis Reagents
Invitrogen™

TaqMan™ Cells-to-CT™ Express Lysis Reagents

This TaqMan Cells-to-CT Express Lysis Reagents product is a convenient, large volume offering of the cell lysis reagents used inRead more
Have Questions?
Catalog NumberQuantity
A579892500 Reactions
Catalog number A57989
Price (INR)
825,593.00
Online offer
Ends: 31-Dec-2026
1,375,987.00
Save 550,394.00 (40%)
Each
Quantity:
2500 Reactions
Price (INR)
825,593.00
Online offer
Ends: 31-Dec-2026
1,375,987.00
Save 550,394.00 (40%)
Each

This TaqMan Cells-to-CT Express Lysis Reagents product is a convenient, large volume offering of the cell lysis reagents used in our TaqMan Cells-to-CT Express Kit. The reagents include Express Lysis Buffer and Express ezDNase. They are ideal for preparing RNA from cultured mammalian cells for gene expression analysis. The lysis procedure simultaneously prepares cell lysates for RT-qPCR and removes genomic DNA in only five minutes. The lysis step is simple to automate with robotic platforms for high-throughput processing of 96- or 384-well plates because it takes place entirely at room temperature. The procedure is also economical as there are only a few pipetting steps and, for cells grown in 96- or 384-well plates, no sample transfers.

Features of the TaqMan Cells-to-CT Express Lysis Reagents include:

  • Extraordinary ease and speed—96 samples ready for RT-qPCR in as little as 5 minutes
  • No tedious RNA purification—no columns, heating, or centrifugation
  • Environmentally friendly—the kit is REACH Compliant, has fewer components, and requires fewer pipetting steps, saving plastics costs
  • Right sized kits for automation—available 2500 reaction size includes both lysis and RT components

Optimized chemistry with no need for stop solution

Gene expression analysis from direct cell lysis is made simple with our TaqMan Cells-to-CT Express Kit. With only one step for cell lysis and no need for the addition of a Stop Solution, our Express technology enables a rapid workflow with sensitive, accurate, and reproducible results. 10–100,000 cells are lysed using the Express Lysis Solution. The optional inclusion of ezDNase to the Express Lysis Solution enables removal of genomic DNA during cell lysis. Following a 5-minute incubation at room temperature, the lysate is used directly for reverse transcription and qPCR analysis.

For Research Use Only.
Specifications
For Use With (Application)Gene Expression
High-throughput CompatibilityYes
Quantity2500 Reactions
Shipping ConditionFrozen, Dry Ice
Isolation TechnologyDirect Cell Lysis
Sample TypeCultured Mammalian Cells
Unit SizeEach
Contents & Storage
• Express Lysis Solution (room temperature or 4°C)
• Express ezDNase (–20°C)

Frequently asked questions (FAQs)

Can I use the Cells-to-CT products with organoids?

Yes. For organoid applications, we recommend the Cells-to-CT Fast Advanced Kit. This kit has been evaluated for direct gene expression analysis from organoid samples and is supported by application data demonstrating its performance.

How many freeze-thaw cycles can the Express Lysis Solution tolerate?

The Express Lysis Solution included in the TaqMan Cells-to-CT Express 1-Step Kit has been tested through at least five freeze-thaw cycles without loss of performance. For laboratories that use the reagent frequently, the Express Lysis Solution can be stored at room temperature or 2-8 degrees C for up to one year.

Would the direct lysis approach to DNA/RNA analysis be a good alternative for very small samples if using the TaqMan Cells-to-CT Express Kit?

Yes. Amplification of RNA targets directly from lysates using the TaqMan Cells-to-CT Express Kit consistently produces better results (lower Ct values) in side-by-side comparisons with column-purified RNA for low sample inputs. For very low sample inputs (fewer than 10 cells), we recommend using Single Cell-to-CT qRT-PCR Kit (Cat. No. 4458236, 4458237).

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

Why don't I need a Stop Solution in the TaqMan Cells-to-CT Express Kit?

The Express Lysis Solution included in the TaqMan Cells-to-CT Express Kit has been reformulated with components that do not require the use of a Stop Solution and yet provide the same level of performance as existing Cells-to-CT kits.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

Can I use the TaqMan Cells-to-CT Express Kit on uncultured primary cells?

This has not been tested. We recommend performing the Pilot Experiment explained in the product manual to determine overall compatibility, maximum input of cells without inhibition, and minimum input of cells for gene detection.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.