TrueMark™ Enteric Bacterial Select Panel I Combo Kit
Applied Biosystems™

TrueMark™ Enteric Bacterial Select Panel I Combo Kit

The TrueMark Enteric Bacterial Select Panel I Combo Kit includes the TrueMark Enteric Bacterial Select Panel I and TrueMark InfectiousRead more
Have Questions?
Catalog NumberQuantity
A58035200 reactions
Catalog number A58035
Price (INR)
408,741.00
Each
Quantity:
200 reactions
Price (INR)
408,741.00
Each

The TrueMark Enteric Bacterial Select Panel I Combo Kit includes the TrueMark Enteric Bacterial Select Panel I and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering. Together with the TrueMark Enteric Bacterial Select Amplification Control I, the kit can be used to quickly evaluate and research up to 200 samples and uses extracted nucleic acid from stool specimens. The kit is recommended for use with the QuantStudio 5 Real-Time PCR System (with 96-well, 0.2-mL block). It includes primers and probe sets specific to the following targets:
Salmonella spp.
Shigella spp./enteroinvasive E. coli (EIEC)
Campylobacter jejuni, Campylobacter coli, Campylobacter upsaliensis
• Shiga toxin-producing E. coli (STEC) stx1/stx2
Bacillus atrophaeus (used as a process control)

Features of the TrueMark Enteric Bacterial Select Panel I Combo Kit include:
• Detects and differentiates between targets found in enteric bacterial infections
• Affordable and scalable—add to existing infrastructure to expand research menu while maintaining low operational costs
• Ready-to-use off-the-shelf solution—seamlessly integrate test into the existing workflow

The TrueMark Enteric Bacterial Select Panel I Combo Kit includes the following components:
TrueMark Enteric Bacterial Select Panel I—multiplexed assay that contains primer and probe sets
TrueMark Infectious Disease 1‐Step Multiplex Master Mix (No ROX)—ready-to-use PCR mix, including polymerase, dNTPs, salt, and buffer

Use this kit with TrueMark Enteric Bacterial Select Amplification Control I, which is a plasmid control containing target-specific templates.
 

Specifications
No. of Reactions200 Reactions
Quantity200 reactions
Detection MethodqPCR
For Use With (Application)Pathogen Detection
FormLiquid
PCR MethodqPCR
Reaction Speed120 min.
Unit SizeEach