TrueMark™ Enteric Bacterial Select Amplification Control I
Applied Biosystems™

TrueMark™ Enteric Bacterial Select Amplification Control I

The TrueMark Enteric Bacterial Select Amplification Control I contains a linearized multi-target plasmid with the following targets: Campylobacter jejuni, Campylobacter coli, Campylobacter upsaliensis, Salmonella spp., Shigella spp./enteroinvasive E. coli (EIEC), Shiga toxin-producing E. coli (STEC) stx1/stx2, and Bacillus atrophaeus.
Have Questions?
Catalog NumberQuantity
A5804925 μL
Catalog number A58049
Price (INR)
99,753.00
Each
Quantity:
25 μL
Price (INR)
99,753.00
Each

The TrueMark Enteric Bacterial Select Amplification Control I contains a linearized multi-target plasmid with the following targets: Campylobacter jejuni, Campylobacter coli, Campylobacter upsaliensis, Salmonella spp., Shigella spp./enteroinvasive E. coli (EIEC), Shiga toxin-producing E. coli (STEC) stx1/stx2, and Bacillus atrophaeus.

The amplification control has a concentration of 1 x 105 copies/μL. It can be used as a positive control for TrueMark Enteric Bacterial Select Panel I-specific amplification or as a stand-alone sample to verify assay performance and help with troubleshooting.

Specifications
Quantity25 μL
Volume25 μL
Detection MethodPrimer-probe
For Use With (Application)Pathogen Detection
FormLiquid
PCR MethodqPCR
Reaction Speed120 min.
Unit SizeEach