TrueMark™ Enteric Bacterial Select Amplification Control II
Applied Biosystems™

TrueMark™ Enteric Bacterial Select Amplification Control II

The TrueMark Enteric Bacterial Select Amplification Control II contains a linearized multi-target plasmid for various targets.
Have Questions?
Catalog NumberQuantity
A5813925 μL
Catalog number A58139
Price (INR)
99,753.00
Each
Quantity:
25 μL
Price (INR)
99,753.00
Each

The TrueMark Enteric Bacterial Select Amplification Control II contains a linearized multi-target plasmid for the following targets: Vibrio (V. parahaemolyticus, V. vulnificus, and V. cholerae), Yersinia enterocolitica, Plesiomonas shigelloides, Aeromonas (A. hydrophila, A. caviae, A. dhakensis), and Bacillus atrophaeus.

This amplification control has a concentration of 1 x 105 copies/µL. It can be used as a positive control for TrueMark Enteric Bacterial Select Panel II-specific amplification or as a stand-alone sample to verify assay performance and help with troubleshooting.

For Research Use Only.
Specifications
Quantity25 μL
Volume25 μL
Detection MethodqPCR
For Use With (Application)Pathogen Detection, Bacterial Enteric Diagnostic Testing, Real Time PCR (qPCR), RT-PCR
FormLiquid
PCR MethodMultiplex qPCR, qPCR
Reaction Speed120 min.
Unit SizeEach