TrueMark™ Enteric Parasitic Select Amplification Control
Applied Biosystems™

TrueMark™ Enteric Parasitic Select Amplification Control

The TrueMark Enteric Parasitic Select Amplification Control contains a linearized multi-target plasmid for the following targets: Cryptosporidium spp., Entamoeba histolytica,Read more
Have Questions?
Catalog NumberQuantity
A5814025 μL
Catalog number A58140
Price (INR)
99,753.00
Each
Quantity:
25 μL
Price (INR)
99,753.00
Each

The TrueMark Enteric Parasitic Select Amplification Control contains a linearized multi-target plasmid for the following targets: Cryptosporidium spp., Entamoeba histolytica, Giardia lamblia, Cyclospora cayetanensis, and Bacillus atrophaeus.

This amplification control has a plasmid concentration of 1 x 105 copies/µL. It can be used as a positive control for TrueMark Enteric Parasitic Select Panel-specific amplification or as a stand-alone sample to verify assay performance and help with troubleshooting.

For Research Use Only.
Specifications
Quantity25 μL
Volume25 μL
Detection MethodqPCR
For Use With (Application)Pathogen Detection, Real Time PCR (qPCR), RT-PCR
FormLiquid
PCR MethodMultiplex qPCR, qPCR, RT-PCR
Reaction Speed120 min.
Unit SizeEach