Pharma Analytics KingFisher™ Flex 96 Deep-Well Tip Combs
Thermo Scientific™
Pharma Analytics KingFisher™ Flex 96 Deep-Well Tip Combs
These tip combs are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Deep-Well Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits.
Have Questions?
Catalog NumberQuantity
A58326PCase of 1
Catalog number A58326P
Price (INR)
54,615.00
線上優惠
Ends: 30-Sep-2026
99,300.00
Save 44,685.00 (45%)
Case of 1
Quantity:
Case of 1
Price (INR)
54,615.00
線上優惠
Ends: 30-Sep-2026
99,300.00
Save 44,685.00 (45%)
Case of 1
These tip combs are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Deep-Well Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits.
Optimize performance using KingFisher Flex System consumables designed specifically for use with these instruments. KingFisher Flex disposable plastics (tip combs and microplates) are made of polypropylene and are ideal for magnetic particle processing because of their low binding affinity for biomolecules. Their special design enables excellent recovery of magnetic beads.
Specifications
DisposableSingle-use
QuantityCase of 1
Format96-well Comb
Product TypeTip Comb
Sample TypeTissue Homogenate
Unit SizeCase of 1