Pharma Analytics KingFisher™ Apex 96 Deep-Well Plates, Barcoded
Thermo Scientific™
Pharma Analytics KingFisher™ Apex 96 Deep-Well Plates, Barcoded
These deep-well plates are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Deep-Well Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits.
Have Questions?
Catalog NumberQuantity
A58358BPCase of 50
Catalog number A58358BP
Price (INR)
47,298.00
Case of 50
Quantity:
Case of 50
Price (INR)
47,298.00
Case of 50
These deep-well plates are for use with the Pharma KingFisher Flex 96 Deep-Well Head Magnetic Particle Processor or MagMAX Express-96 Deep-Well Magnetic Particle Processor in combination with PrepSEQ ResDNA kits and ProteinSEQ kits. The deep-well configuration differs from the standard configuration in that the volume of sample input is from 100 to 1,000 μL per well compared to 20–200 μL for the standard configuration.
Optimize performance using KingFisher Flex System consumables designed specifically for use with these instruments. KingFisher Flex disposable plastics (tip combs and microplates) are made of polypropylene and are ideal for magnetic particle processing because of their low binding affinity for biomolecules. Their special design enables excellent recovery of magnetic beads.
Specifications
AutoclavableNot Autoclavable
DisposableSingle-use
LidWithout Lid
Product TypeDeep-Well Plate
QuantityCase of 50
Skirt StyleNon-skirt
ColorClear
Fit for PurposeCertified Quality
MaterialPolypropylene
No. of Wells96
Product LineKingFisher
SterilityNon-sterile
Surface TreatmentLow Binding
Well ShapeConical Bottom (V-well)
Unit SizeCase of 50