NaCl Solutions
Gibco™

NaCl Solutions

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentration
A59067BD100 L5 M
A59067BA5 L5 M
A59067BB20 L5 M
A59067BC50 L5 M
A59067BE200 L5 M
A65781BA5 L4 M
A65781BB20 L4 M
A65781BC50 L4 M
A65781BD100 L4 M
A65781BE200 L4 M
A65779BA5 L3 M
A65779BB20 L3 M
A65779BC50 L3 M
A65779BD100 L3 M
A65779BE200 L3 M
Catalog number A59067BD
Price (INR)
-
Quantity:
100 L
Concentration:
5 M

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.

Features

  • Concentrated for dilution on-site
  • Available in a range of pack sizes
  • Established shelf life: set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification

Benefits

  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages

Additional information

  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: TAegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS7647-14-5
Chemical Name or MaterialRaw Material: Sodium Chloride
Manufacturing QualityCGMP
PackagingBioprocess Container in Secondary Packaging
Concentration5 M
For Use With (Application)Bioproduction
For Use With (Equipment)ILC/ILD System
GradeEP/USP
Molecular Weight (g/mol)Raw Material: 58.44
Percent PurityNot Available
Physical FormSolution
Quantity100 L
Solution TypeBuffer
pH3.0 to 10.0
Unit SizeEach