DPBS, no calcium, no magnesium
Gibco™

DPBS, no calcium, no magnesium

Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A59095BC50 L
A59095BA5 L
A59095BB20 L
A59095BD100 L
A59095BE200 L
Catalog number A59095BC
Price (INR)
-
Quantity:
50 L
Gibco™ Balanced Salt Solutions are sterile-filtered, packaged in bioprocess containers, and ready to order. Commonly used for processing cells and flushing equipment, sourcing Gibco balanced salt solutions can help streamline your operation by reducing buffer preparation and allowing you to focus on additional critical steps in your biopharmaceutical processes. Discover new outsourcing efficiencies to reclaim time, space, and resources.

Product Features

  • Multiple formulations and concentrations available
  • Available in a range of pack sizes (5 L, 20 L, 50 L, 100 L, and 200 L)
  • Established products with set specifications are made to order
  • Product standardization enables shorter times from inquiry to quote and shipment, compared to custom
  • Readily accessible documentation for accelerated qualification

Benefits

  • Reduces time spent preparing buffers in-house
  • Mitigates space requirement for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Meets workflow needs across stages of development and manufacturing of biologicals
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialDPBS
ColorClear, colorless solution
IncludesNo Calcium, No Magnesium
Manufacturing QualitycGMP
Osmolality270 to 300 mOsm/kg
PackagingDrum
SterilitySterile-filtered
GradeNot Available
Physical FormLiquid
Quantity50 L
pH7.0 to 7.3
Unit SizeEach