Tris 20 mM, NaCl 1.5 M
Tris 20 mM, NaCl 1.5 M
Gibco™
Tris 20 mM, NaCl 1.5 M
Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A59168BD100 L
A59168BA5 L
A59168BB20 L
A59168BC50 L
A59168BE200 L
Catalog number A59168BD
Price (INR)
-
Request A Quote
Quantity:
100 L
Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.
Features
  • Available in a range of pack sizes
  • Established shelf life; set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification
Benefits
  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages
Additional information
  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: Aegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
FormulationTris/NaCl Buffer
Manufacturing QualityCGMP
Quantity100 L
FormClear, colorless solution
Product TypeSodium Chloride
SterilitySterile-filtered
Sterilization MethodSterile-filtered
Unit SizeEach