CTS™ Rotea™ Full Flow Single-Use Kit
CTS™ Rotea™ Full Flow Single-Use Kit
Gibco™

CTS™ Rotea™ Full Flow Single-Use Kit

The CTS Rotea Single-Use Kit is part of the CTS Rotea Counterflow Centrifugation System. The kit has been validated to operate within the full system flow rate range of 6–150 mL/min.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A5917210 Pk.
A591735 Pk.
Catalog number A59172
Price (INR)
-
Request A Quote
Quantity:
10 Pk.

The Gibco CTS Rotea Full Flow Single-Use Kit is part of the CTS Rotea Counterflow Centrifugation System. The kit has been validated to operate within the full system flow rate range of 6–150 mL/min. It can be used to perform a wide range of cell processing steps, including wash & concentrate, separation, isolation, and more. The single-use kit provides flexibility for process development and easily supports commercial-scale manufacturing. The kit is compatible with sterile luer connectors or thermal welding.

Tubing material: DEHP-free PVC

Tube dimensions: 4 mm OD, 2.4 mm ID

Pump tube material: PharmaPure

Kits are gamma-irradiated and individually packaged.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypeAgnostic
Certifications/ComplianceISO 13485
DisposableSingle-use
Flow Rate6 to 150 mL/min.
For Use With (Application)Cell and Gene Therapy Research, Development, and Manufacturing
MaterialTritan MX711 Copolyester (Chamber), DEHP-free PVC (Tubing)
Max. RCF3000 x g
Quantity10 Pk.
Shelf Life12 months from date of manufacture
Sterilization MethodGamma-irradiated
For Use With (Equipment)CTS Rotea Counterflow Centrifugation System
Unit Size10 kits