RapidLINK™ Software v2.0
Applied Biosystems™
RapidLINK™ Software v2.0
The Applied Biosystems RapidLINK Software v2.0 is part of the Applied Biosystems RapidHIT ID System.
Have Questions?
Catalog NumberQuantity
A59183Ea.
Catalog number A59183
Price (INR)
1,407,747.00
Each
Quantity:
Ea.
Price (INR)
1,407,747.00
Each
The Applied Biosystems RapidLINK Software v2.0 is part of the Applied Biosystems RapidHIT ID System. This fast and simple DNA system produces lab-quality forensic DNA profiles that are aggregated within the software for direct upload to the database of choice or for further review and analysis. This data management software expedites identity confirmation securely and efficiently.
Features of the RapidLINK Software v2.0 include:
  • Securely manage one or more Applied Biosystems RapidHIT ID systems from a central location>
  • Review DNA profile results in real time prior to upload to a database
  • Enable or disable users in real time
  • Collect and review metadata using built-in reporting tools
  • Expert system-capable
  • Sample/data accessioning workflow-capable
Delivery method: After purchase you will receive a letter with upgrade/installation details.
For Research Use Only.
Specifications
Product LineRapidLINK
Product TypeSoftware
QuantityEa.
Shipping ConditionRoom Temperature
Unit SizeEach