CTS™ Cellmation™ Software for DeltaV™ Systems
CTS™ Cellmation™ Software for DeltaV™ Systems
CTS™ Cellmation™ Software for DeltaV™ Systems
CTS™ Cellmation™ Software for DeltaV™ Systems
Gibco™
CTS™ Cellmation™ Software for DeltaV™ Systems
CTS Cellmation Software is an off-the-shelf solution designed to help simplify and optimize cell therapy manufacturing processes by providing digital connectivity for modular cell therapy instrumentation offered by Thermo Fisher Scientific.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesQuantity
A59196Full Library5 Modules
A59197Incubator Module1 Module
A59198CryoMed Module1 Module
Catalog number A59196
Price (INR)
-
Includes:
Full Library
Quantity:
5 Modules
Gibco CTS Cellmation Software is an off-the-shelf solution designed to help simplify and optimize cell therapy manufacturing processes by providing digital connectivity for modular cell therapy instrumentation offered by Thermo Fisher Scientific. The software’s open platform provides a flexible system for managing and integrating multiple instruments, streamlining the cell therapy workflow and helping ensure regulatory compliance while minimizing cost and time.
CTS Cellmation Software offers the following benefits:
  • Off-the-shelf solution—removes the need for costly custom software projects and extensive validation, helping save time and resources in the manufacturing process
  • Seamless integration—connect and control your cell therapy workflow across multiple instruments through a single, user-friendly interface
  • Data security and compliance using DeltaV Live software—help ensure traceable, reproducible, and secure data storage with the DeltaV Continuous Historian, helping enable compliance with the requirements 21 CFR Part 11/Annex 11 for cGMP
  • Streamlined data management—direct connection to the Emerson DeltaV DCS layer simplifies the transfer of data streams to manufacturing execution system (MES) and enterprise resource planning (ERP) systems
  • Workflow control—create user-specific batch recipes to control the flow of your cell therapy manufacturing process
  • Ongoing support—service contracts provide both on-site and remote support; installation is performed by Thermo Fisher Scientific representatives with remote support offered 12 hours a day, 5 days a week
CTS Cellmation Software is powered by the DeltaV DCS (Distributed Control System) from Emerson and has been developed following GAMP5 methods to help ensure compatibility with cGMP-compliant processes. CTS Cellmation Software is optimized to easily scale from process development through clinical manufacturing and commercialization.
For Research Use or Manufacturing of Cell, Gene, or Tissue- Based Products.
Specifications
Certifications/ComplianceSupports 21 CFR part 11/Annex 11 compliance
DescriptionFull Library
For Use With (Equipment)Emerson's DeltaV Distributed Control System (DCS), CTS™ DynaCellect™ Magnetic Separation System, CTS™ Rotea™ Counterflow Centrifugation System, CTS™ Xenon™ Electroporation System, CryoMed™ Controlled-Rate Freezer, Heracell™ Vios™ CR CO2 Incubator
IncludesFull Library
Operating SystemDeltaV Live v14.LTS
Quantity5 Modules
Unit SizeEach