VetMAX™ African Swine Fever Virus MGF360-CD2V DIVA Controls
Applied Biosystems™

VetMAX™ African Swine Fever Virus MGF360-CD2V DIVA Controls

The VetMAX African Swine Fever Virus MGF360-CD2V DIVA Controls are synthetic positive controls used in the real-time PCR amplification of CD2V and MGF360-14L genes of African swine fever virus.
Have Questions?
Catalog NumberQuantity
A59803500 Reactions
Catalog number A59803
Price (INR)
-
Quantity:
500 Reactions

The VetMAX African Swine Fever Virus MGF360-CD2V DIVA Controls are synthetic positive controls used in the real-time PCR amplification of CD2V and MGF360-14L genes of African swine fever virus.

For Research Use Only.
Specifications
DiseaseAfrican Swine Fever
No. of Reactions500 reactions
Product TypeControls
Quantity500 Reactions
Detection MethodqPCR
For Use With (Application)Pathogen Detection, Virus Detection
FormLiquid
GC-Rich PCR PerformanceMedium
Label or DyeUnlabeled
PCR MethodqPCR
Reaction Speed30 to 50 min.
Unit Size500 reactions