VetMAX™ PRRSV NA High Concentration Controls
Applied Biosystems™

VetMAX™ PRRSV NA High Concentration Controls

VetMAX NA High Concentration Controls are for use in reverse-transcription real-time PCR (RT-qPCR) workflows targeting the North American strain of porcine reproductive and respiratory syndrome virus (PRRSV).
Have Questions?
Catalog NumberQuantity
A65256200 μL
Catalog number A65256
Price (INR)
18,612.00
Each
Quantity:
200 μL
Price (INR)
18,612.00
Each

VetMAX PRRSV NA High Concentration Controls are for use in reverse-transcription real-time PCR (RT-qPCR) workflows targeting the North American strain of porcine reproductive and respiratory syndrome virus (PRRSV). The controls are for use with VetMAX PRRSV 3.0 products in all the applications where standards are required (e.g., quantitative PCR, validation tests).

Notes

  • Contains only PRRSV NA high concentration control
  • No Xeno included
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DiseasePRRSV
No. of ReactionsVaries by dilution
Product TypeControls
Quantity200 μL
Shelf Life12 Months
Shipping ConditionShipped at -10°C to -30°C
SpeciesPorcine
Volume200 μL
Concentration1 x 106 Copies/μL
Detection MethodqPCR, RT-PCR
For Use With (Application)Real Time PCR (qPCR)
FormLiquid
GC-Rich PCR PerformanceMedium
PCR MethodRT-qPCR
Reaction SpeedFast or Standard
Unit SizeEach