PrioCHECK™ Paratuberculosis Ab 3.0 Plate Kit
PrioCHECK™ Paratuberculosis Ab 3.0 Plate Kit
PrioCHECK™ Paratuberculosis Ab 3.0 Plate Kit
PrioCHECK™ Paratuberculosis Ab 3.0 Plate Kit
Applied Biosystems™

PrioCHECK™ Paratuberculosis Ab 3.0 Plate Kit

The Applied Biosystems PrioCHECK Paratuberculosis Ab 3.0 Plate Kit is an easy‐to‐use ELISA that enables detection of antibodies against Mycobacterium avium ssp. paratuberculosis (MAP) in samples from ruminants (serum, heparinized plasma, or milk).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySufficient For
A6541630 plates, 2760 tests2760 Tests
A654155 plates, 460 tests460 Tests
Catalog number A65416
Price (INR)
-
Quantity:
30 plates, 2760 tests
Sufficient For:
2760 Tests

The Applied Biosystems PrioCHECK Paratuberculosis Ab 3.0 Plate Kit is an easy‐to‐use ELISA that enables detection of antibodies against Mycobacterium avium ssp. paratuberculosis (MAP) in samples from ruminants (serum, heparinized plasma, or milk).

Features of the PrioCHECK Paratuberculosis Ab 3.0 Plate Kit include:

  • Room temperature shipping
  • Universal protocol that works across all ruminant species including bovine, ovine, and caprine
  • Flexible protocols with short and long incubation options

Paratuberculosis, also known as Johne's Disease, results from an infection by MAP, a gram-positive bacillus. This infection primarily targets the mesenteric ganglia and small intestine, leading to a chronic intestinal infection. Clinical symptoms include diarrhea, weight loss, and edema. These symptoms become noticeable only in the advanced stages of the disease. Disease control relies on identifying infected animals and executing culling procedures.

Regulatory requirements vary by country; products may not be available in your geographic area.
Specifications
AssayELISA
Product LinePrioCHECK
Quantity30 plates, 2760 tests
Regulatory StatusFor Veterinary Use Only. For In Vitro Use Only.
Shipping ConditionRoom Temperature
Sufficient For2760 Tests
TargetMycobacterium avium ssp. Paratuberculosis (MAP)
Target Species ValidatedBovine, Goat, Sheep
TechniqueIndirect ELISA
Immunoassay Kit FormatStrip Plate
Sample TypeSerum, Heparin Plasma, Milk
Target SpeciesMulti-species, Bovine, Caprine, Ovine
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C