POROS™ CaptureSelect™ C1-InhXL Affinity Resin
Thermo Scientific™

POROS™ CaptureSelect™ C1-InhXL Affinity Resin

Thermo Scientific™ POROS™ CaptureSelect™ C1-InhXL Affinity Resin is intended for the purification of human C1 esterase inhibitor from recombinant and plasma/serum sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A654415 L
A6543625 mL
A6543750 mL
A65439250 mL
A654401 L
A6544210 L
Catalog number A65441
Price (INR)
-
Quantity:
5 L

POROS CaptureSelect C1-InhXL Affinity Resin is intended for the purification of human C1 esterase inhibitor from recombinant and plasma/serum sources. C1 esterase inhibitor is a protease inhibitor belonging to the serpin superfamily that inhibits the spontaneous activation of the complement system. The CaptureSelect C1-InhXL affinity ligand is highly selective for human C1 inhibitor and coupled to the high-performing POROS resin, allowing for an efficient and scalable purification process, with high purity and yield obtained in a single step. The large pore structure of the POROS beads enables higher flowrates and efficient binding of larger biomolecules.

Features of POROS CaptureSelect C1-InhXL Affinity Resin include:

  • Efficient purification of C1 esterase inhibitor with high yield
  • Mild pH elution condition to retain activity
  • High binding capacity at high flow rates
  • Excellent scalability
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Resin: cross-linked poly(styrene-divinylbenzene) POROS beads
  • Average particle size: 50 μm
  • Ligand: single-domain [VHH] antibody fragment
  • Binding capacity: 20–25 g C1-Inhibitor/L resin
  • Elution conditions: 50 mM sodium acetate, 1.0 M MgCl2, pH 5.0
  • Regeneration conditions: acidic stripping condition with 0.5 M acetic acid, pH 2
  • Mechanical resistance: 100 bar (1450 psi, 10 MPa)
  • Formulation buffer: 18% (v/v) ethanol
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Binding Capacity20 to 25 g C1-Inhibitor/L resin
For Use With (Application)Protein purification
Quantity5 L
Shipping ConditionWet Ice
TargetC1 Esterase Inhibitor
FormResin Slurry
Particle Size50 μm
Product LinePOROS CaptureSelect
TypeAffinity Resin
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.