KingFisher™ PlasmidPro Maxi Processor, extended warranty package
KingFisher™ PlasmidPro Maxi Processor, extended warranty package
KingFisher™ PlasmidPro Maxi Processor, extended warranty package
KingFisher™ PlasmidPro Maxi Processor, extended warranty package
Thermo Scientific™

KingFisher™ PlasmidPro Maxi Processor, extended warranty package

The KingFisher PlasmidPro Maxi Processor transforms the maxi-scale plasmid purification process with a cutting-edge, fully automated workflow that eliminates labor-intensiveRead more
Have Questions?
Catalog NumberQuantity
A664271 each
Catalog number A66427
Price (INR)
-
Quantity:
1 each

The KingFisher PlasmidPro Maxi Processor transforms the maxi-scale plasmid purification process with a cutting-edge, fully automated workflow that eliminates labor-intensive centrifugation and pipetting steps. Simply pour in the culture and let the instrument handle the rest. In under 75 minutes, this innovative system efficiently executes all essential purification steps, delivering high-quality, endotoxin-free plasmid DNA.

True walkaway automation

The KingFisher PlasmidPro system is fully automated, eliminating the need for pipetting or centrifugation steps. It completes each run in under 75 minutes, allowing you to focus on other tasks.

Exceptional quality

Optimized for maxi-scale plasmid DNA purification from 150 mL of bacterial culture, the system consistently delivers high-quality, advanced, transfection-grade plasmid DNA.

Convenient pre-filled cartridge

The one-piece, pre-filled KingFisher PlasmidPro Maxi Processor Endotoxin-Free Cartridge eliminates setup time and ensures a simple clean-up process, enabling quick turnaround for your next purification.

Effortless installation and setup

The installation process can be completed in minutes on any benchtop, ensuring immediate usability.

AB Assurance Plan

The AB Assurance Plan is a comprehensive repair plan that aims to optimize instrument performance and ensure the availability of critical systems. It includes preventive maintenance, proactive instrument monitoring, and repairs, all aimed at keeping your laboratory operating smoothly.

Additional details

The KingFisher PlasmidPro Maxi Processor is specifically optimized for cultures grown in Luria-Bertani (LB) medium.

At the end of the procedure, the system delivers endotoxin-free plasmid DNA resuspended in TE buffer, ensuring optimal quality and compatibility with downstream applications.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dimensions19.7 x 11.8 x 23.6 in. (50 x 30 x 60 cm)
Display7 in. Touchscreen
Electrical Requirements100-240 VAC, 50/60 Hz (nominal), automatic voltage detection
For Use With (Application)Plasmid purification, advanced transfection applications
Memory∼30 GB
PortsUSB
Purification TargetPlasmid DNA
Quantity1 each
Run Time<75 min.
Sample Volume150 mL
Samples per Run1 sample/run
Shipping ConditionRoom temperature
FormatCartridge
Sample TypeBacterial Culture
Unit SizeEach

Frequently asked questions (FAQs)

Can I resuspend a frozen pellet for use with the KingFisher PlasmidPro Maxi Processor?

When using frozen pellets as a starting point, please consider the following guidelines:

  • The pellet should represent 150 mL of the specified culture.
  • Fully resuspend the pellet in 150 mL of 1X PBS or the parent media.
  • The resuspension can be achieved by either pipetting up and down or vigorous vortexing.


Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

What are the storage conditions for the KingFisher PlasmidPro Maxi Processor Cartridge? Is refrigeration required?

We recommend storing the KingFisher PlasmidPro Maxi Processor Cartridge at room temperature. Refrigeration is not needed.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

Can I reuse the KingFisher PlasmidPro Maxi Processor Cartridge?

The KingFisher PlasmidPro Maxi Processor Cartridge is single-use and should be disposed of after use, minimizing clean-up and risk of cross-contamination. Re-use of the cartridge will contaminate and potentially harm the instrument.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

Is the KingFisher PlasmidPro Maxi Processor Cartridge animal origin-free?

No. The RNaseA used in the KingFisher PlasmidPro Maxi Processor Cartridge is not animal origin-free. Review the Certificate of Analysis (COA) for more details.

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.

What solution is the plasmid DNA sample resuspended in at the end of the purification using the KingFisher PlasmidPro Maxi Processor?

At the end of the purification using the KingFisher PlasmidPro Maxi Processor, the plasmid DNA sample is resuspended in TE buffer (10 mM Tris-HCl, pH 8.0, 0.1 mM EDTA).

Find additional tips, troubleshooting help, and resources within our Automated Nucleic Acid Purification Support Center.