Product Image
Applied Biosystems™
HID Real-Time PCR Analysis Software v1.4 Upgrade
HID Real-Time PCR Analysis Software is a tool that helps enable forensic laboratories to optimize their quantification workflow and enhance their downstream decision-making process.
Have Questions?
Catalog NumberQuantity
A789021 Ea.
Catalog number A78902
Price (INR)
-
Quantity:
1 Ea.
HID Real-Time PCR Analysis Software is a tool that helps enable forensic laboratories to optimize their quantification workflow and enhance their downstream decision-making process.
The software has the following workflow features:
  • Compatible with firmware v1.5.1 of QuantStudio 5 Real-Time PCR System for Human Identification
  • Virtual standard curve functionality
  • Customizable templates
  • Advanced data analysis capabilities including quality flags
  • STR sample normalization (dilution) and reaction setup tool
HID Real-Time PCR Analysis Software v1.4 is designed for use with the QuantStudio 5 real-time PCR systems running firmware v1.5.1. It offers a set of features beyond instrument controls. HID Real-Time PCR Analysis Software v1.4 streamlines the experimental setup, supports customizable workflow templates, and enhances the data analysis and review process. It is validated for use on the Windows™ 10 platform and it may be used with all available HID Quantifiler kits.
This product includes an HID Real-Time PCR Analysis Software v1.4 Upgrade DVD and 5 user licenses to upgrade from v1.2 of the software. Documentation is not included.
Specifications
For Use With (Equipment)QuantStudio 5 Real-Time PCR Systems running firmware v1.5.1
Product TypeHID Real-Time PCR Analysis Software v1.4 Upgrade
Quantity1 Ea.
Shipping ConditionRoom Temperature
SoftwareReal-Time PCR Analysis
Unit SizeEach