Difco™ TC Yeastolate, ultra-filtered (UF)
Gibco™

Difco™ TC Yeastolate, ultra-filtered (UF)

Enhance cell culture performance with Gibco™ Difco™ TC Yeastolate Ultra-Filtered (UF) Low Bioburden while also mitigating risk in bioprocessing methods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityManufacturing Quality
A80503-0110 kgUF, Low Bioburden (≤300 CFU)
292804500 gUF
29280510 kgUF
67007950 kgUF
A80501-0110 kgUF, Low Bioburden (≤100 CFU)
A80501-0250 kgUF, Low Bioburden (≤100 CFU)
A80503-0250 kgUF, Low Bioburden (≤300 CFU)
Catalog number A80503-01
Price (INR)
-
Quantity:
10 kg
Manufacturing Quality:
UF, Low Bioburden (≤300 CFU)
Difco TC Yeastolate UF Low Bioburden is an animal origin-free and water-soluble extract of autolyzed yeast, enhanced for tissue culture (TC) applications. This product is a mixture of low molecular weight peptides, amino acids, carbohydrates (simple and complex), and vitamins. In addition to the performance-enhancing features of our TC yeastolate product, this offering can help reduce endotoxin and bioburden levels.

Difco TC Yeastolate, ultra-filtered (UF), is an animal origin-free (AOF) product derived from water-soluble portions of autolyzed yeast (Saccharomyces cerevisiae). This product is intended as a nutritional supplement for bacterial, insect, and mammalian cell culture.

Low bioburden
• Reducing bioburden levels limits the effects of potential interactions between the organisms and the peptone, which could impact product stability and performance
• May eliminate the need to remove higher levels of bioburden from incoming critical raw materials, therefore helping to improve the upstream bioproduction process
• Available in bioburden levels of ≤100–300 CFU

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineCHO, HEK, Sf9, MDCK, BHK, Vero, High Five
Cell TypeMammalian, Insect, Microbial
ColorBeige
Culture TypeMammalian, Insect, Microbial
For Use With (Application)Preparing bacterial, insect, and mammalian cell culture media
FormatPail
FormulationPowder with pH 5.4 to 7.2
Manufacturing QualityUF, Low Bioburden (≤300 CFU)
Quantity10 kg
Serum LevelSerum-free
Shelf Life48 Months
ClassificationAnimal Origin-free
FormPowder
Product TypeTC Yeastolate
SterilityNon-sterile
Without AdditivesNo Animal Derived Components
Unit Size10 kg