Sample Storage Freezer Racks
Product Image
Thermo Scientific™
Sample Storage Freezer Racks
Maximize compact storage of microplates, blocks and tube racks in a cold storage unit or on the benchtop with Thermo Scientific™ Sample Storage Freezer Racks.
Have Questions?
Catalog NumberHolds
AB-1031G
also known as AB-1031/G
Boxes (34 mm)
Catalog number AB-1031G
also known as AB-1031/G
Price (INR)
-
Request A Quote
Holds:
Boxes (34 mm)
Maximize compact storage of microplates, blocks and tube racks in a cold storage unit or on the benchtop with Thermo Scientific™ Sample Storage Freezer Racks. Our team has a strong reputation for supporting both equipment and consumables, and can determine which sample storage freezer rack is best for your laboratory storage needs.
Versatile Design
  • Thermo Scientific sample storage freezer racks are available for use with microplates, blocks and storage tube racks up to 70mm tall
  • Twin positioned handles facilitate use in both chest and upright freezers and contents are kept in position during transportation with a locking rod
Secure Storage
  • Maximize bench top space or enable compact storage in your refrigerator or freezer down to -80°C with our freezer racks
Trusted Resource
  • Researchers worldwide protect more than two billion samples inside Thermo Scientific cold storage products
  • With proven equipment including cryopreservation units, storage tubes, capping systems and barcode readers, you're free to concentrate on your work without worrying about your valuable samples
Specifications
Description34mm
HoldsBoxes (34 mm)
Product TypeFreezer Rack
Unit SizeEach