ThermoPrime <i>Taq</i> DNA Polymerase, with 10X buffer and separate vial of 25 mM MgCl<sub>2</sub>
Thermo Scientific™

ThermoPrime Taq DNA Polymerase, with 10X buffer and separate vial of 25 mM MgCl2

Thermo Scientific ThermoPrime Taq Taq Polymerase is a 94 kDa recombinant thermostable DNA polymerase obtained by high level expression of the Taq DNA polymerase gene in E. coli.
Have Questions?
Catalog NumberQuantity
AB0301B2500 units
Catalog number AB0301B
Price (INR)
127,178.00
Online offer
Ends: 31-Dec-2026
211,963.00
Save 84,785.00 (40%)
Each
Quantity:
2500 units
Price (INR)
127,178.00
Online offer
Ends: 31-Dec-2026
211,963.00
Save 84,785.00 (40%)
Each
Thermo Scientific ThermoPrimeTaq Polymerase is a 94-kDa recombinant thermostable DNA polymerase obtained by high level expression of the Taq DNA Polymerase gene in E. coli. A proprietary buffer component protects ThermoPrime DNA Polymerase during high temperature steps, ensuring optimal performance throughout the reaction, leading to increased PCR yields. Convenient master mix format is also available for easier reaction set-up.

Features

  • Enhanced enzyme stability for increased yield
  • Advanced buffering system minimizes PCR optimization steps

Applications

  • Amplification of fragments up to 5 kb
  • Routine PCR
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)1X
Hot StartNo
No. of Reactions4000 Reactions
Overhang3'-A
PolymeraseThermoPrime Taq DNA Polymerase
Product TypeDNA Polymerase
Quantity2500 units
Reaction FormatSeparate Components
Shipping ConditionWet or Dry Ice
Starting MaterialDNA
Concentration5 U/μL
For Use With (Application)Standard PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• ThermoPrime Taq DNA Polymerase (5 U/μL), 10 x 50 μL
• 10X Reaction Buffer IV, 10 x 1.5 mL
• 25 mM MgCl2, 10 x 1.5 mL

Store at -20°C.